Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36261553-36261804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOL1
Entrez Gene ID
8542
Gene Name
apolipoprotein L1
Gene Aliases
APO-L, APOL, APOL-I, FSGS4
UniGene
Hs.114309
Gene Chromosome Location
Chr.22: 36253071-36267531 on Build GRCh38
Transcripts
NM_001136540.1 NM_001136541.1 NM_003661.3 NM_145343.2 XM_005261796.3 XM_011530478.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4797022 c.255G>A p.E85E Chr.22: 36261615-36261615 on Build GRCh38
4400135 c.253G>A p.E85K Chr.22: 36261613-36261613 on Build GRCh38
1183269 c.323A>G p.E108G Chr.22: 36261683-36261683 on Build GRCh38
4836741 c.322G>T p.E108* Chr.22: 36261682-36261682 on Build GRCh38
4797023 c.395G>A p.R132K Chr.22: 36261615-36261615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041205676 Chr.22: 36261723-36261723 on Build GRCh38
rs758230408 Chr.22: 36261765-36261765 on Build GRCh38
rs748124806 Chr.22: 36261761-36261761 on Build GRCh38
rs778033694 Chr.22: 36261593-36261593 on Build GRCh38
rs774981170 Chr.22: 36261750-36261750 on Build GRCh38
rs751841122 Chr.22: 36261680-36261680 on Build GRCh38
rs749213200 Chr.22: 36261627-36261627 on Build GRCh38
rs779899888 Chr.22: 36261620-36261620 on Build GRCh38
rs781432998 Chr.22: 36261612-36261612 on Build GRCh38
rs136164 Chr.22: 36261743-36261743 on Build GRCh38
rs570768141 Chr.22: 36261581-36261581 on Build GRCh38
rs757708122 Chr.22: 36261681-36261681 on Build GRCh38
rs767714990 Chr.22: 36261683-36261683 on Build GRCh38
rs776102873 Chr.22: 36261663-36261663 on Build GRCh38
rs200109088 Chr.22: 36261666-36261666 on Build GRCh38
rs761706641 Chr.22: 36261643-36261643 on Build GRCh38
rs764713348 Chr.22: 36261674-36261674 on Build GRCh38
rs201153239 Chr.22: 36261603-36261603 on Build GRCh38
rs112074559 Chr.22: 36261693-36261693 on Build GRCh38
rs776453961 Chr.22: 36261769-36261772 on Build GRCh38
rs370836972 Chr.22: 36261758-36261758 on Build GRCh38
rs969926809 Chr.22: 36261619-36261619 on Build GRCh38
rs779109637 Chr.22: 36261747-36261747 on Build GRCh38
rs764504559 Chr.22: 36261745-36261745 on Build GRCh38
rs370874898 Chr.22: 36261736-36261736 on Build GRCh38
rs750592758 Chr.22: 36261755-36261755 on Build GRCh38
rs747878269 Chr.22: 36261735-36261735 on Build GRCh38
rs746693537 Chr.22: 36261741-36261741 on Build GRCh38
rs559819422 Chr.22: 36261719-36261719 on Build GRCh38
rs754837072 Chr.22: 36261705-36261705 on Build GRCh38
rs1005971808 Chr.22: 36261584-36261584 on Build GRCh38
rs758725190 Chr.22: 36261591-36261591 on Build GRCh38
rs745944908 Chr.22: 36261617-36261617 on Build GRCh38
rs780128413 Chr.22: 36261697-36261697 on Build GRCh38
rs746853150 Chr.22: 36261774-36261774 on Build GRCh38
rs41297245 Chr.22: 36261694-36261694 on Build GRCh38
rs12106612 Chr.22: 36261757-36261757 on Build GRCh38
rs955704200 Chr.22: 36261700-36261700 on Build GRCh38
rs920228875 Chr.22: 36261622-36261622 on Build GRCh38
rs1047514407 Chr.22: 36261661-36261661 on Build GRCh38
rs766243964 Chr.22: 36261733-36261747 on Build GRCh38
rs777737416 Chr.22: 36261772-36261772 on Build GRCh38
rs771615220 Chr.22: 36261650-36261650 on Build GRCh38
rs768448236 Chr.22: 36261630-36261630 on Build GRCh38
rs755141458 Chr.22: 36261760-36261760 on Build GRCh38
rs752984894 Chr.22: 36261658-36261658 on Build GRCh38
rs928958548 Chr.22: 36261657-36261657 on Build GRCh38
rs136163 Chr.22: 36261582-36261582 on Build GRCh38
rs759127522 Chr.22: 36261744-36261744 on Build GRCh38
rs141871473 Chr.22: 36261768-36261768 on Build GRCh38
rs749432798 Chr.22: 36261699-36261699 on Build GRCh38
rs751756905 Chr.22: 36261598-36261598 on Build GRCh38
rs376102138 Chr.22: 36261728-36261728 on Build GRCh38
rs372444766 Chr.22: 36261637-36261637 on Build GRCh38
rs762318604 Chr.22: 36261588-36261588 on Build GRCh38
rs767899707 Chr.22: 36261754-36261754 on Build GRCh38

View Full Product Details