Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3751505-3751740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBA3
Entrez Gene ID
9546
Gene Name
amyloid beta precursor protein binding family A member 3
Gene Aliases
MGC:15815, X11L2, mint3
UniGene
Hs.25527
Gene Chromosome Location
Chr.19: 3750773-3761697 on Build GRCh38
Transcripts
NM_004886.3 XM_006722950.3 XM_006722951.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1612131 c.1401G>T p.T467T Chr.19: 3751548-3751548 on Build GRCh38
6601876 c.1412C>T p.T471M Chr.19: 3751537-3751537 on Build GRCh38
439336 c.1401G>A p.T467T Chr.19: 3751548-3751548 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769775292 Chr.19: 3751548-3751548 on Build GRCh38
rs889414556 Chr.19: 3751532-3751532 on Build GRCh38
rs746656418 Chr.19: 3751536-3751536 on Build GRCh38
rs775632915 Chr.19: 3751563-3751563 on Build GRCh38
rs774725531 Chr.19: 3751608-3751608 on Build GRCh38
rs774134080 Chr.19: 3751558-3751558 on Build GRCh38
rs760814326 Chr.19: 3751569-3751569 on Build GRCh38
rs754594706 Chr.19: 3751537-3751537 on Build GRCh38
rs897275007 Chr.19: 3751716-3751716 on Build GRCh38
rs183105670 Chr.19: 3751595-3751595 on Build GRCh38
rs954995342 Chr.19: 3751611-3751611 on Build GRCh38
rs911501038 Chr.19: 3751622-3751622 on Build GRCh38
rs930065455 Chr.19: 3751682-3751682 on Build GRCh38
rs938727365 Chr.19: 3751712-3751712 on Build GRCh38
rs548806609 Chr.19: 3751665-3751665 on Build GRCh38
rs980205888 Chr.19: 3751687-3751687 on Build GRCh38
rs1019296459 Chr.19: 3751534-3751534 on Build GRCh38
rs752192985 Chr.19: 3751593-3751593 on Build GRCh38
rs528885772 Chr.19: 3751540-3751540 on Build GRCh38
rs974302165 Chr.19: 3751649-3751649 on Build GRCh38
rs754612089 Chr.19: 3751590-3751590 on Build GRCh38
rs775452454 Chr.19: 3751564-3751564 on Build GRCh38
rs765702851 Chr.19: 3751574-3751574 on Build GRCh38
rs777742054 Chr.19: 3751605-3751605 on Build GRCh38
rs565806902 Chr.19: 3751603-3751603 on Build GRCh38
rs141575653 Chr.19: 3751527-3751527 on Build GRCh38
rs770235603 Chr.19: 3751573-3751573 on Build GRCh38
rs759680039 Chr.19: 3751562-3751562 on Build GRCh38
rs747648736 Chr.19: 3751543-3751543 on Build GRCh38
rs986857837 Chr.19: 3751613-3751613 on Build GRCh38
rs928653426 Chr.19: 3751704-3751705 on Build GRCh38
rs773106862 Chr.19: 3751549-3751549 on Build GRCh38
rs764738525 Chr.19: 3751570-3751570 on Build GRCh38
rs773822465 Chr.19: 3751580-3751581 on Build GRCh38
rs749144277 Chr.19: 3751551-3751551 on Build GRCh38
rs770695052 Chr.19: 3751552-3751552 on Build GRCh38
rs750762177 Chr.19: 3751589-3751589 on Build GRCh38
rs948678387 Chr.19: 3751651-3751651 on Build GRCh38
rs989364231 Chr.19: 3751599-3751599 on Build GRCh38
rs931588845 Chr.19: 3751719-3751719 on Build GRCh38
rs560491204 Chr.19: 3751696-3751696 on Build GRCh38
rs780600772 Chr.19: 3751542-3751542 on Build GRCh38
rs938881835 Chr.19: 3751700-3751700 on Build GRCh38
rs112086597 Chr.19: 3751533-3751533 on Build GRCh38
rs754383819 Chr.19: 3751571-3751571 on Build GRCh38
rs1037199727 Chr.19: 3751709-3751709 on Build GRCh38
rs914590344 Chr.19: 3751640-3751640 on Build GRCh38

View Full Product Details