Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 111142218-111142491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L11
Entrez Gene ID
10018
Gene Name
BCL2 like 11
Gene Aliases
BAM, BIM, BOD
UniGene
Hs.469658
Gene Chromosome Location
Chr.2: 111120914-111168445 on Build GRCh38
Transcripts
NM_001204106.1 NM_001204107.1 NM_001204108.1 NM_001204109.1 NM_001204110.1 NM_001204111.1 NM_001204112.1 NM_001204113.1 NM_006538.4 NM_138621.4 NM_138622.3 NM_138623.3 NM_138624.3 NM_138625.3 NM_138626.3 NM_138627.3 NM_207003.2 XM_005263550.3 XM_005263551.2 XM_005263552.3 XM_005263553.2 XM_005263554.2 XM_005263555.3 XM_005263556.3 XM_005263557.4 XM_005263559.2 XM_005263561.3 XM_017003101.1 XM_017003102.1

SNP Detail

SNP ID Chromosome Location
rs1025421099 Chr.2: 111142351-111142351 on Build GRCh38
rs201667919 Chr.2: 111142309-111142309 on Build GRCh38
rs548910377 Chr.2: 111142296-111142297 on Build GRCh38
rs547430589 Chr.2: 111142317-111142317 on Build GRCh38
rs3761706 Chr.2: 111142304-111142304 on Build GRCh38
rs1037866625 Chr.2: 111142321-111142321 on Build GRCh38
rs933556491 Chr.2: 111142290-111142290 on Build GRCh38
rs565420761 Chr.2: 111142434-111142434 on Build GRCh38
rs749268046 Chr.2: 111142391-111142391 on Build GRCh38
rs374980122 Chr.2: 111142384-111142384 on Build GRCh38
rs188068366 Chr.2: 111142431-111142431 on Build GRCh38
rs770871307 Chr.2: 111142400-111142400 on Build GRCh38
rs923281245 Chr.2: 111142281-111142281 on Build GRCh38
rs1033601403 Chr.2: 111142453-111142453 on Build GRCh38
rs994303965 Chr.2: 111142337-111142337 on Build GRCh38
rs148743335 Chr.2: 111142320-111142320 on Build GRCh38
rs946150095 Chr.2: 111142259-111142259 on Build GRCh38
rs114223633 Chr.2: 111142368-111142368 on Build GRCh38
rs538359859 Chr.2: 111142302-111142302 on Build GRCh38
rs200453151 Chr.2: 111142377-111142377 on Build GRCh38
rs567082237 Chr.2: 111142404-111142405 on Build GRCh38
rs1018105916 Chr.2: 111142388-111142388 on Build GRCh38
rs574204935 Chr.2: 111142263-111142263 on Build GRCh38
rs977428731 Chr.2: 111142269-111142269 on Build GRCh38

View Full Product Details