Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 43882516-43882787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH3
Entrez Gene ID
221120
Gene Name
alkB homolog 3, alpha-ketoglutaratedependent dioxygenase
Gene Aliases
ABH3, DEPC-1, DEPC1, PCA1, hABH3
UniGene
Hs.720708
Gene Chromosome Location
Chr.11: 43880807-43920275 on Build GRCh38
Transcripts
NM_139178.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4661143 c.17G>A p.R6Q Chr.11: 43882669-43882669 on Build GRCh38
4641463 c.46G>A p.A16T Chr.11: 43882698-43882698 on Build GRCh38
4753991 c.52G>A p.V18I Chr.11: 43882704-43882704 on Build GRCh38
5882073 c.79+2C>T p.? Chr.11: 43882733-43882733 on Build GRCh38
926915 c.12A>C p.K4N Chr.11: 43882664-43882664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778860310 Chr.11: 43882693-43882693 on Build GRCh38
rs766880377 Chr.11: 43882640-43882640 on Build GRCh38
rs578010060 Chr.11: 43882598-43882598 on Build GRCh38
rs749077175 Chr.11: 43882675-43882675 on Build GRCh38
rs760784166 Chr.11: 43882610-43882610 on Build GRCh38
rs748441294 Chr.11: 43882733-43882733 on Build GRCh38
rs145974652 Chr.11: 43882666-43882666 on Build GRCh38
rs754439335 Chr.11: 43882653-43882653 on Build GRCh38
rs368061935 Chr.11: 43882678-43882678 on Build GRCh38
rs747116638 Chr.11: 43882716-43882716 on Build GRCh38
rs777430638 Chr.11: 43882659-43882660 on Build GRCh38
rs542570191 Chr.11: 43882651-43882651 on Build GRCh38
rs1043393522 Chr.11: 43882600-43882600 on Build GRCh38
rs778429024 Chr.11: 43882740-43882740 on Build GRCh38
rs776952533 Chr.11: 43882625-43882625 on Build GRCh38
rs1040688650 Chr.11: 43882564-43882564 on Build GRCh38
rs199576340 Chr.11: 43882672-43882672 on Build GRCh38
rs766821611 Chr.11: 43882742-43882742 on Build GRCh38
rs769021867 Chr.11: 43882619-43882619 on Build GRCh38
rs148553066 Chr.11: 43882671-43882671 on Build GRCh38
rs774885140 Chr.11: 43882743-43882743 on Build GRCh38
rs896645015 Chr.11: 43882692-43882692 on Build GRCh38
rs757019324 Chr.11: 43882752-43882753 on Build GRCh38
rs752196063 Chr.11: 43882757-43882757 on Build GRCh38
rs201182770 Chr.11: 43882633-43882633 on Build GRCh38
rs907099847 Chr.11: 43882627-43882627 on Build GRCh38
rs575548759 Chr.11: 43882741-43882741 on Build GRCh38
rs765641506 Chr.11: 43882629-43882629 on Build GRCh38
rs767069098 Chr.11: 43882755-43882755 on Build GRCh38
rs553343513 Chr.11: 43882555-43882555 on Build GRCh38
rs1021754405 Chr.11: 43882684-43882684 on Build GRCh38
rs746755417 Chr.11: 43882739-43882739 on Build GRCh38
rs759057079 Chr.11: 43882753-43882753 on Build GRCh38
rs762274113 Chr.11: 43882628-43882628 on Build GRCh38
rs770138186 Chr.11: 43882734-43882734 on Build GRCh38
rs913830266 Chr.11: 43882581-43882582 on Build GRCh38
rs371627856 Chr.11: 43882731-43882731 on Build GRCh38
rs752284114 Chr.11: 43882668-43882668 on Build GRCh38
rs562893856 Chr.11: 43882751-43882752 on Build GRCh38
rs755748430 Chr.11: 43882669-43882669 on Build GRCh38
rs773492606 Chr.11: 43882735-43882735 on Build GRCh38
rs377089877 Chr.11: 43882736-43882736 on Build GRCh38
rs773613793 Chr.11: 43882632-43882632 on Build GRCh38

View Full Product Details