Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 118239754-118240023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD7
Entrez Gene ID
56311
Gene Name
ankyrin repeat domain 7
Gene Aliases
TSA806
UniGene
Hs.729340
Gene Chromosome Location
Chr.7: 118224658-118242731 on Build GRCh38
Transcripts
NM_019644.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1084511 c.776G>T p.R259I Chr.7: 118239912-118239912 on Build GRCh38
4988282 c.803G>A p.G268E Chr.7: 118239939-118239939 on Build GRCh38
4506781 c.789C>T p.F263F Chr.7: 118239925-118239925 on Build GRCh38
1084512 c.795G>A p.A265A Chr.7: 118239931-118239931 on Build GRCh38
4609286 c.789C>A p.F263L Chr.7: 118239925-118239925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767500490 Chr.7: 118239930-118239930 on Build GRCh38
rs769878439 Chr.7: 118239851-118239851 on Build GRCh38
rs748469940 Chr.7: 118239963-118239963 on Build GRCh38
rs778492926 Chr.7: 118239994-118239994 on Build GRCh38
rs1028719864 Chr.7: 118239823-118239823 on Build GRCh38
rs189882379 Chr.7: 118239914-118239914 on Build GRCh38
rs759179242 Chr.7: 118239874-118239874 on Build GRCh38
rs576496837 Chr.7: 118239953-118239953 on Build GRCh38
rs757199806 Chr.7: 118239922-118239922 on Build GRCh38
rs772554991 Chr.7: 118239993-118239993 on Build GRCh38
rs772640742 Chr.7: 118239870-118239870 on Build GRCh38
rs185347467 Chr.7: 118239852-118239852 on Build GRCh38
rs774727137 Chr.7: 118239812-118239812 on Build GRCh38
rs937921070 Chr.7: 118239800-118239800 on Build GRCh38
rs200629473 Chr.7: 118239931-118239931 on Build GRCh38
rs756225177 Chr.7: 118239934-118239934 on Build GRCh38
rs12154434 Chr.7: 118239786-118239786 on Build GRCh38
rs367864583 Chr.7: 118239873-118239873 on Build GRCh38
rs368645249 Chr.7: 118239977-118239977 on Build GRCh38
rs930479924 Chr.7: 118239938-118239938 on Build GRCh38
rs373637370 Chr.7: 118239892-118239892 on Build GRCh38
rs973331369 Chr.7: 118239846-118239846 on Build GRCh38
rs114353258 Chr.7: 118239893-118239893 on Build GRCh38
rs764838715 Chr.7: 118239890-118239890 on Build GRCh38
rs143223202 Chr.7: 118239861-118239861 on Build GRCh38
rs779150403 Chr.7: 118239962-118239962 on Build GRCh38
rs545972486 Chr.7: 118239799-118239799 on Build GRCh38
rs749496386 Chr.7: 118239952-118239952 on Build GRCh38
rs780161800 Chr.7: 118239945-118239945 on Build GRCh38
rs147870989 Chr.7: 118239905-118239905 on Build GRCh38

View Full Product Details