Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151117292-151117547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGAP3
Entrez Gene ID
116988
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
Gene Aliases
AGAP-3, CENTG3, CRAG, MRIP-1, cnt-g3
UniGene
Hs.647075
Gene Chromosome Location
Chr.7: 151085831-151144436 on Build GRCh38
Transcripts
NM_001042535.3 NM_001281300.1 NM_001308304.1 NM_001308305.1 NM_031946.6 XM_005249943.2 XM_011515780.1 XM_017011732.1 XM_017011733.1 XM_017011734.1 XM_017011735.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3636322 c.552C>T p.P184P Chr.7: 151117444-151117444 on Build GRCh38
3636320 c.538G>A p.D180N Chr.7: 151117430-151117430 on Build GRCh38
3636323 c.552C>T p.P184P Chr.7: 151117444-151117444 on Build GRCh38
3261608 c.549delC p.P185fs*74 Chr.7: 151117441-151117441 on Build GRCh38
5762553 c.479-2A>G p.? Chr.7: 151117369-151117369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778257654 Chr.7: 151117473-151117473 on Build GRCh38
rs1017735384 Chr.7: 151117494-151117494 on Build GRCh38
rs927230619 Chr.7: 151117340-151117340 on Build GRCh38
rs192463556 Chr.7: 151117522-151117522 on Build GRCh38
rs548513717 Chr.7: 151117322-151117322 on Build GRCh38
rs374551644 Chr.7: 151117357-151117357 on Build GRCh38
rs777585300 Chr.7: 151117443-151117443 on Build GRCh38
rs993878869 Chr.7: 151117393-151117393 on Build GRCh38
rs887845649 Chr.7: 151117481-151117481 on Build GRCh38
rs768695091 Chr.7: 151117471-151117471 on Build GRCh38
rs200290690 Chr.7: 151117356-151117356 on Build GRCh38
rs762350944 Chr.7: 151117355-151117355 on Build GRCh38
rs759301175 Chr.7: 151117501-151117501 on Build GRCh38
rs753462151 Chr.7: 151117359-151117359 on Build GRCh38
rs758833571 Chr.7: 151117370-151117370 on Build GRCh38
rs956137401 Chr.7: 151117369-151117369 on Build GRCh38
rs768748451 Chr.7: 151117328-151117328 on Build GRCh38
rs988748491 Chr.7: 151117441-151117441 on Build GRCh38
rs774821074 Chr.7: 151117330-151117330 on Build GRCh38
rs367550525 Chr.7: 151117476-151117476 on Build GRCh38
rs770001050 Chr.7: 151117320-151117320 on Build GRCh38
rs771529709 Chr.7: 151117378-151117378 on Build GRCh38
rs758476423 Chr.7: 151117409-151117409 on Build GRCh38
rs762299421 Chr.7: 151117478-151117478 on Build GRCh38
rs372558355 Chr.7: 151117488-151117488 on Build GRCh38
rs749563182 Chr.7: 151117469-151117469 on Build GRCh38
rs372739605 Chr.7: 151117339-151117339 on Build GRCh38
rs527748673 Chr.7: 151117408-151117408 on Build GRCh38
rs547595660 Chr.7: 151117468-151117468 on Build GRCh38
rs371386280 Chr.7: 151117487-151117487 on Build GRCh38
rs539634505 Chr.7: 151117485-151117485 on Build GRCh38
rs367961665 Chr.7: 151117495-151117495 on Build GRCh38
rs965201211 Chr.7: 151117520-151117520 on Build GRCh38
rs776632154 Chr.7: 151117358-151117358 on Build GRCh38
rs746753299 Chr.7: 151117464-151117464 on Build GRCh38
rs188703168 Chr.7: 151117489-151117489 on Build GRCh38

View Full Product Details