Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 134918646-134918913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH8A1
Entrez Gene ID
64577
Gene Name
aldehyde dehydrogenase 8 family member A1
Gene Aliases
ALDH12, DJ352A20.2
UniGene
Hs.486520
Gene Chromosome Location
Chr.6: 134917390-134950122 on Build GRCh38
Transcripts
NM_001193480.1 NM_022568.3 NM_170771.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5939485 c.1201C>T p.P401S Chr.6: 134918678-134918678 on Build GRCh38
3948066 c.1128G>T p.T376T Chr.6: 134918751-134918751 on Build GRCh38
740018 c.1084A>T p.S362C Chr.6: 134918795-134918795 on Build GRCh38
4761720 c.1018A>C p.S340R Chr.6: 134918861-134918861 on Build GRCh38
5869357 c.1031G>A p.R344K Chr.6: 134918848-134918848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753503411 Chr.6: 134918693-134918693 on Build GRCh38
rs755497194 Chr.6: 134918856-134918857 on Build GRCh38
rs184006801 Chr.6: 134918706-134918706 on Build GRCh38
rs147929376 Chr.6: 134918856-134918856 on Build GRCh38
rs148657521 Chr.6: 134918872-134918872 on Build GRCh38
rs769819663 Chr.6: 134918727-134918727 on Build GRCh38
rs150799987 Chr.6: 134918689-134918689 on Build GRCh38
rs373644944 Chr.6: 134918747-134918747 on Build GRCh38
rs140383315 Chr.6: 134918816-134918816 on Build GRCh38
rs916000443 Chr.6: 134918805-134918805 on Build GRCh38
rs935225695 Chr.6: 134918870-134918870 on Build GRCh38
rs61731731 Chr.6: 134918678-134918678 on Build GRCh38
rs774508271 Chr.6: 134918754-134918754 on Build GRCh38
rs141798638 Chr.6: 134918682-134918682 on Build GRCh38
rs928794896 Chr.6: 134918786-134918786 on Build GRCh38
rs760654383 Chr.6: 134918797-134918797 on Build GRCh38
rs144907106 Chr.6: 134918804-134918804 on Build GRCh38
rs767279629 Chr.6: 134918777-134918777 on Build GRCh38
rs774809603 Chr.6: 134918752-134918752 on Build GRCh38
rs575696584 Chr.6: 134918853-134918853 on Build GRCh38
rs2294315 Chr.6: 134918674-134918674 on Build GRCh38
rs749408208 Chr.6: 134918871-134918871 on Build GRCh38
rs901664830 Chr.6: 134918885-134918885 on Build GRCh38
rs536504709 Chr.6: 134918688-134918688 on Build GRCh38
rs149883779 Chr.6: 134918803-134918803 on Build GRCh38
rs772181224 Chr.6: 134918719-134918719 on Build GRCh38
rs756650191 Chr.6: 134918860-134918860 on Build GRCh38
rs778084962 Chr.6: 134918869-134918869 on Build GRCh38
rs759367815 Chr.6: 134918762-134918762 on Build GRCh38
rs75706536 Chr.6: 134918855-134918855 on Build GRCh38
rs747404906 Chr.6: 134918713-134918713 on Build GRCh38
rs781773753 Chr.6: 134918718-134918718 on Build GRCh38
rs370890664 Chr.6: 134918717-134918717 on Build GRCh38
rs766231142 Chr.6: 134918712-134918712 on Build GRCh38
rs751359207 Chr.6: 134918681-134918681 on Build GRCh38
rs752686912 Chr.6: 134918785-134918785 on Build GRCh38
rs748639830 Chr.6: 134918725-134918725 on Build GRCh38
rs138997999 Chr.6: 134918751-134918751 on Build GRCh38
rs750072643 Chr.6: 134918830-134918830 on Build GRCh38
rs756669749 Chr.6: 134918814-134918814 on Build GRCh38

View Full Product Details