The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152331.3 | NP_689544.3 | 3 - 3 | 1179 | 88 |
GenBank mRNA | AK055797.1 | - | 3 - 3 | 725 | 88 |
AK094223.1 | - | 3 - 3 | 948 | 88 | |
BC031799.1 | - | 2 - 2 | 338 | 88 | |
BC090945.1 | - | 3 - 3 | 1179 | 88 | |
BC117341.1 | - | 3 - 3 | 958 | 88 | |
BC117343.1 | - | 3 - 3 | 958 | 88 | |
BX248047.1 | - | 3 - 3 | 718 | 88 |