Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23561879-23562138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1G2
Entrez Gene ID
8906
Gene Name
adaptor related protein complex 1 gamma 2 subunit
Gene Aliases
G2AD
UniGene
Hs.343244
Gene Chromosome Location
Chr.14: 23559563-23569056 on Build GRCh38
Transcripts
NM_001282474.1 NM_001282475.1 NM_003917.4 XM_005268167.3 XM_005268168.4 XM_005268169.3 XM_005268170.3 XM_005268172.3 XM_005268173.3 XM_005268174.3 XM_005268175.3 XM_005268177.3 XM_005268178.3 XM_005268179.3 XM_005268180.4 XM_005268182.3 XM_006720301.3 XM_011537283.2 XM_011537286.2 XM_017021740.1 XM_017021741.1 XM_017021742.1 XM_017021746.1 XM_017021747.1 XM_017021748.1 NR_110555.1

Gene Symbol
LOC102724814
Entrez Gene ID
102724814
Gene Name
uncharacterized LOC102724814
Gene Aliases
-
UniGene
Hs.569375
Gene Chromosome Location
Chr.14: 23561097-23568076 on Build GRCh38
Transcripts
NM_001282474.1 NM_001282475.1 NM_003917.4 XM_005268167.3 XM_005268168.4 XM_005268169.3 XM_005268170.3 XM_005268172.3 XM_005268173.3 XM_005268174.3 XM_005268175.3 XM_005268177.3 XM_005268178.3 XM_005268179.3 XM_005268180.4 XM_005268182.3 XM_006720301.3 XM_011537283.2 XM_011537286.2 XM_017021740.1 XM_017021741.1 XM_017021742.1 XM_017021746.1 XM_017021747.1 XM_017021748.1 NR_110555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6412926 c.1283A>C p.K428T Chr.14: 23561977-23561977 on Build GRCh38
6428766 c.1241A>C p.E414A Chr.14: 23562019-23562019 on Build GRCh38
5478661 c.1202G>A p.R401H Chr.14: 23562058-23562058 on Build GRCh38
6382506 c.1643T>C p.V548A Chr.14: 23562052-23562052 on Build GRCh38
5478660 c.1637G>A p.R546H Chr.14: 23562058-23562058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368747270 Chr.14: 23562039-23562039 on Build GRCh38
rs148282907 Chr.14: 23562059-23562059 on Build GRCh38
rs148525847 Chr.14: 23562008-23562008 on Build GRCh38
rs754938216 Chr.14: 23562003-23562003 on Build GRCh38
rs890597421 Chr.14: 23561999-23561999 on Build GRCh38
rs576634833 Chr.14: 23562067-23562067 on Build GRCh38
rs769627554 Chr.14: 23561956-23561956 on Build GRCh38
rs758206386 Chr.14: 23561937-23561937 on Build GRCh38
rs373148237 Chr.14: 23561935-23561935 on Build GRCh38
rs753122091 Chr.14: 23561979-23561979 on Build GRCh38
rs779648112 Chr.14: 23562094-23562094 on Build GRCh38
rs747797122 Chr.14: 23561949-23561949 on Build GRCh38
rs760007030 Chr.14: 23561973-23561973 on Build GRCh38
rs199999308 Chr.14: 23562012-23562012 on Build GRCh38
rs952501873 Chr.14: 23562075-23562075 on Build GRCh38
rs778910779 Chr.14: 23561994-23561994 on Build GRCh38
rs369245203 Chr.14: 23561947-23561947 on Build GRCh38
rs978635859 Chr.14: 23562033-23562033 on Build GRCh38
rs773343932 Chr.14: 23561973-23561974 on Build GRCh38
rs202232189 Chr.14: 23562065-23562065 on Build GRCh38
rs367610818 Chr.14: 23561972-23561972 on Build GRCh38
rs948473619 Chr.14: 23562106-23562106 on Build GRCh38
rs768476906 Chr.14: 23561942-23561942 on Build GRCh38
rs754886923 Chr.14: 23562113-23562113 on Build GRCh38
rs371365778 Chr.14: 23561981-23561981 on Build GRCh38
rs771300290 Chr.14: 23561969-23561969 on Build GRCh38
rs746734267 Chr.14: 23561941-23561941 on Build GRCh38
rs773770477 Chr.14: 23562044-23562044 on Build GRCh38
rs765355477 Chr.14: 23562071-23562071 on Build GRCh38
rs761777964 Chr.14: 23561997-23561997 on Build GRCh38
rs748756065 Chr.14: 23562038-23562038 on Build GRCh38
rs767581949 Chr.14: 23561917-23561917 on Build GRCh38
rs760833509 Chr.14: 23561980-23561980 on Build GRCh38
rs777821677 Chr.14: 23561926-23561926 on Build GRCh38
rs764307065 Chr.14: 23561988-23561988 on Build GRCh38
rs934440194 Chr.14: 23561914-23561914 on Build GRCh38
rs756399107 Chr.14: 23561923-23561923 on Build GRCh38
rs767564556 Chr.14: 23562117-23562117 on Build GRCh38
rs76920030 Chr.14: 23561989-23561989 on Build GRCh38
rs369638005 Chr.14: 23561921-23561921 on Build GRCh38
rs772788148 Chr.14: 23561957-23561957 on Build GRCh38
rs377511055 Chr.14: 23561940-23561940 on Build GRCh38
rs375677387 Chr.14: 23562064-23562064 on Build GRCh38
rs200780955 Chr.14: 23562058-23562058 on Build GRCh38
rs762899254 Chr.14: 23562044-23562045 on Build GRCh38
rs751633676 Chr.14: 23561911-23561911 on Build GRCh38
rs376968278 Chr.14: 23562023-23562023 on Build GRCh38
rs149292012 Chr.14: 23561961-23561961 on Build GRCh38
rs781121124 Chr.14: 23562007-23562007 on Build GRCh38
rs924502802 Chr.14: 23562035-23562035 on Build GRCh38
rs777598503 Chr.14: 23562027-23562027 on Build GRCh38
rs750526449 Chr.14: 23562082-23562082 on Build GRCh38
rs746377212 Chr.14: 23562051-23562051 on Build GRCh38
rs767888618 Chr.14: 23561975-23561975 on Build GRCh38
rs374503007 Chr.14: 23562031-23562031 on Build GRCh38
rs553433716 Chr.14: 23561903-23561903 on Build GRCh38

View Full Product Details