Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25307775-25308046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD12
Entrez Gene ID
26090
Gene Name
abhydrolase domain containing 12
Gene Aliases
ABHD12A, BEM46L2, C20orf22, PHARC, dJ965G21.2
UniGene
Hs.441550
Gene Chromosome Location
Chr.20: 25294743-25390982 on Build GRCh38
Transcripts
NM_001042472.2 NM_015600.4 XM_005260698.1 XM_011529214.2 XM_011529215.1 XM_011529218.1 XM_017027796.1 XM_017027798.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1751360 c.866T>G p.V289G Chr.20: 25307967-25307967 on Build GRCh38
4134340 c.837C>T p.R279R Chr.20: 25307996-25307996 on Build GRCh38
6374486 c.799G>A p.D267N Chr.20: 25308034-25308034 on Build GRCh38
1181397 c.865G>A p.V289M Chr.20: 25307968-25307968 on Build GRCh38
1751361 c.866T>G p.V289G Chr.20: 25307967-25307967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191089093 Chr.20: 25307941-25307941 on Build GRCh38
rs773050687 Chr.20: 25307881-25307883 on Build GRCh38
rs139497461 Chr.20: 25308029-25308029 on Build GRCh38
rs397704714 Chr.20: 25307980-25307981 on Build GRCh38
rs980374873 Chr.20: 25307903-25307906 on Build GRCh38
rs770956336 Chr.20: 25307994-25307994 on Build GRCh38
rs6107027 Chr.20: 25307996-25307996 on Build GRCh38
rs748457154 Chr.20: 25307929-25307929 on Build GRCh38
rs372807311 Chr.20: 25307975-25307975 on Build GRCh38
rs994701660 Chr.20: 25307820-25307820 on Build GRCh38
rs571950585 Chr.20: 25307940-25307940 on Build GRCh38
rs1024315955 Chr.20: 25307892-25307892 on Build GRCh38
rs1007497383 Chr.20: 25307905-25307905 on Build GRCh38
rs884613 Chr.20: 25307869-25307869 on Build GRCh38
rs890336290 Chr.20: 25307900-25307900 on Build GRCh38
rs143805685 Chr.20: 25307899-25307899 on Build GRCh38
rs751503495 Chr.20: 25308024-25308024 on Build GRCh38
rs999134336 Chr.20: 25307829-25307829 on Build GRCh38
rs375955961 Chr.20: 25307916-25307916 on Build GRCh38
rs1026382946 Chr.20: 25307867-25307869 on Build GRCh38
rs762636997 Chr.20: 25308014-25308014 on Build GRCh38
rs988959018 Chr.20: 25307993-25307993 on Build GRCh38
rs995580201 Chr.20: 25307995-25307995 on Build GRCh38
rs764133867 Chr.20: 25308016-25308016 on Build GRCh38
rs370259926 Chr.20: 25307923-25307923 on Build GRCh38
rs775417181 Chr.20: 25308007-25308007 on Build GRCh38
rs575909094 Chr.20: 25307803-25307803 on Build GRCh38
rs761432556 Chr.20: 25308028-25308028 on Build GRCh38
rs145964823 Chr.20: 25307998-25307998 on Build GRCh38
rs1018267576 Chr.20: 25307906-25307906 on Build GRCh38
rs759550508 Chr.20: 25307926-25307928 on Build GRCh38
rs180761451 Chr.20: 25307997-25307997 on Build GRCh38

View Full Product Details