Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25006753-25007026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSS1
Entrez Gene ID
84532
Gene Name
acyl-CoA synthetase short-chain family member 1
Gene Aliases
ACAS2L, ACECS1, AceCS2L
UniGene
Hs.529353
Gene Chromosome Location
Chr.20: 25006230-25058182 on Build GRCh38
Transcripts
NM_001252675.1 NM_001252676.1 NM_001252677.1 NM_032501.3 XM_006723659.1 XM_011529388.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5589523 c.1681T>C p.F561L Chr.20: 25006907-25006907 on Build GRCh38
5766614 c.1727A>G p.*576* Chr.20: 25006861-25006861 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs955986113 Chr.20: 25006833-25006833 on Build GRCh38
rs748043979 Chr.20: 25006948-25006948 on Build GRCh38
rs567111480 Chr.20: 25006914-25006914 on Build GRCh38
rs550713864 Chr.20: 25006786-25006786 on Build GRCh38
rs768390192 Chr.20: 25006923-25006923 on Build GRCh38
rs929732620 Chr.20: 25006879-25006879 on Build GRCh38
rs973739798 Chr.20: 25006842-25006842 on Build GRCh38
rs149129309 Chr.20: 25006913-25006913 on Build GRCh38
rs749555334 Chr.20: 25006832-25006832 on Build GRCh38
rs186170584 Chr.20: 25006830-25006830 on Build GRCh38
rs904645314 Chr.20: 25006823-25006823 on Build GRCh38
rs774177849 Chr.20: 25006929-25006929 on Build GRCh38
rs768535821 Chr.20: 25006922-25006922 on Build GRCh38
rs6083718 Chr.20: 25006802-25006802 on Build GRCh38
rs1057291043 Chr.20: 25006776-25006776 on Build GRCh38
rs895999299 Chr.20: 25006779-25006779 on Build GRCh38
rs982606531 Chr.20: 25006903-25006903 on Build GRCh38
rs921106052 Chr.20: 25006934-25006934 on Build GRCh38
rs749258922 Chr.20: 25006909-25006909 on Build GRCh38
rs780073126 Chr.20: 25006826-25006826 on Build GRCh38
rs1023114610 Chr.20: 25006803-25006803 on Build GRCh38
rs919634448 Chr.20: 25006853-25006853 on Build GRCh38

View Full Product Details