Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 12896324-12896595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP44
Entrez Gene ID
9912
Gene Name
Rho GTPase activating protein 44
Gene Aliases
NPC-A-10, RICH2
UniGene
Hs.499758
Gene Chromosome Location
Chr.17: 12789512-12991644 on Build GRCh38
Transcripts
NM_001321164.1 NM_001321166.1 NM_001321167.1 NM_001321168.1 NM_014859.5 NR_135569.1 XM_005256891.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
40228 c.176G>A p.G59E Chr.17: 12896489-12896489 on Build GRCh38
6209770 c.134G>A p.S45N Chr.17: 12896447-12896447 on Build GRCh38
6231112 c.138G>A p.T46T Chr.17: 12896451-12896451 on Build GRCh38
6251548 c.178G>T p.A60S Chr.17: 12896491-12896491 on Build GRCh38
6556193 c.176G>T p.G59V Chr.17: 12896489-12896489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758419215 Chr.17: 12896407-12896407 on Build GRCh38
rs780194876 Chr.17: 12896409-12896409 on Build GRCh38
rs759876429 Chr.17: 12896452-12896452 on Build GRCh38
rs376421836 Chr.17: 12896475-12896475 on Build GRCh38
rs766790521 Chr.17: 12896371-12896371 on Build GRCh38
rs574773463 Chr.17: 12896557-12896557 on Build GRCh38
rs564691983 Chr.17: 12896399-12896399 on Build GRCh38
rs758330878 Chr.17: 12896481-12896481 on Build GRCh38
rs375297564 Chr.17: 12896463-12896463 on Build GRCh38
rs202018989 Chr.17: 12896511-12896511 on Build GRCh38
rs780760718 Chr.17: 12896530-12896530 on Build GRCh38
rs776234269 Chr.17: 12896450-12896450 on Build GRCh38
rs774542227 Chr.17: 12896373-12896373 on Build GRCh38
rs373275512 Chr.17: 12896521-12896521 on Build GRCh38
rs192254681 Chr.17: 12896466-12896466 on Build GRCh38
rs894683478 Chr.17: 12896518-12896518 on Build GRCh38
rs756252171 Chr.17: 12896386-12896386 on Build GRCh38
rs73978273 Chr.17: 12896382-12896382 on Build GRCh38
rs768335427 Chr.17: 12896446-12896446 on Build GRCh38
rs915193050 Chr.17: 12896392-12896392 on Build GRCh38
rs367607261 Chr.17: 12896417-12896417 on Build GRCh38
rs374981240 Chr.17: 12896415-12896415 on Build GRCh38
rs889111381 Chr.17: 12896454-12896454 on Build GRCh38
rs767864905 Chr.17: 12896459-12896459 on Build GRCh38
rs377403306 Chr.17: 12896552-12896552 on Build GRCh38
rs778796877 Chr.17: 12896391-12896391 on Build GRCh38
rs776467977 Chr.17: 12896362-12896362 on Build GRCh38
rs746916340 Chr.17: 12896425-12896425 on Build GRCh38
rs148457740 Chr.17: 12896560-12896560 on Build GRCh38
rs202156982 Chr.17: 12896370-12896370 on Build GRCh38
rs753820014 Chr.17: 12896467-12896467 on Build GRCh38
rs754137391 Chr.17: 12896390-12896390 on Build GRCh38
rs768264041 Chr.17: 12896361-12896361 on Build GRCh38
rs779976295 Chr.17: 12896483-12896483 on Build GRCh38
rs62637651 Chr.17: 12896451-12896451 on Build GRCh38
rs747555699 Chr.17: 12896548-12896548 on Build GRCh38
rs756226920 Chr.17: 12896389-12896389 on Build GRCh38
rs1046007169 Chr.17: 12896571-12896571 on Build GRCh38
rs748908544 Chr.17: 12896561-12896561 on Build GRCh38
rs919148759 Chr.17: 12896448-12896448 on Build GRCh38
rs767956724 Chr.17: 12896381-12896381 on Build GRCh38

View Full Product Details