Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18184220-18184469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH5
Entrez Gene ID
54890
Gene Name
alkB homolog 5, RNA demethylase
Gene Aliases
ABH5, OFOXD, OFOXD1
UniGene
Hs.744130
Gene Chromosome Location
Chr.17: 18183553-18209954 on Build GRCh38
Transcripts
NM_017758.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3667960 c.177G>A p.K59K Chr.17: 18184420-18184420 on Build GRCh38
401684 c.212A>T p.Y71F Chr.17: 18184455-18184455 on Build GRCh38
3387731 c.135C>T p.A45A Chr.17: 18184378-18184378 on Build GRCh38
1191096 c.98_99insCGCCGC p.A37_V38insAA Chr.17: 18184341-18184342 on Build GRCh38
3889390 c.154C>T p.P52S Chr.17: 18184397-18184397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769720160 Chr.17: 18184404-18184404 on Build GRCh38
rs950334125 Chr.17: 18184271-18184271 on Build GRCh38
rs759019429 Chr.17: 18184435-18184435 on Build GRCh38
rs748081999 Chr.17: 18184401-18184401 on Build GRCh38
rs768757008 Chr.17: 18184385-18184385 on Build GRCh38
rs780246148 Chr.17: 18184382-18184382 on Build GRCh38
rs143278496 Chr.17: 18184238-18184238 on Build GRCh38
rs771220641 Chr.17: 18184361-18184366 on Build GRCh38
rs370188460 Chr.17: 18184441-18184441 on Build GRCh38
rs766201317 Chr.17: 18184417-18184417 on Build GRCh38
rs747095824 Chr.17: 18184384-18184384 on Build GRCh38
rs998121179 Chr.17: 18184381-18184381 on Build GRCh38
rs1014963993 Chr.17: 18184374-18184374 on Build GRCh38
rs757591442 Chr.17: 18184306-18184306 on Build GRCh38
rs1016845020 Chr.17: 18184313-18184313 on Build GRCh38
rs764965117 Chr.17: 18184339-18184356 on Build GRCh38
rs750394889 Chr.17: 18184340-18184348 on Build GRCh38
rs950930378 Chr.17: 18184288-18184288 on Build GRCh38
rs887898438 Chr.17: 18184356-18184356 on Build GRCh38
rs758673141 Chr.17: 18184377-18184377 on Build GRCh38
rs764549752 Chr.17: 18184264-18184264 on Build GRCh38
rs746393020 Chr.17: 18184373-18184381 on Build GRCh38
rs961548571 Chr.17: 18184378-18184378 on Build GRCh38
rs746103468 Chr.17: 18184360-18184360 on Build GRCh38
rs61999283 Chr.17: 18184442-18184442 on Build GRCh38
rs1020111860 Chr.17: 18184347-18184348 on Build GRCh38
rs754503234 Chr.17: 18184356-18184357 on Build GRCh38
rs912907074 Chr.17: 18184450-18184450 on Build GRCh38
rs965449817 Chr.17: 18184358-18184381 on Build GRCh38
rs1016527303 Chr.17: 18184379-18184379 on Build GRCh38
rs779180587 Chr.17: 18184345-18184345 on Build GRCh38
rs761566390 Chr.17: 18184334-18184342 on Build GRCh38
rs762828500 Chr.17: 18184414-18184414 on Build GRCh38
rs772765591 Chr.17: 18184372-18184373 on Build GRCh38
rs779550198 Chr.17: 18184343-18184366 on Build GRCh38
rs751084337 Chr.17: 18184348-18184356 on Build GRCh38
rs773087086 Chr.17: 18184405-18184405 on Build GRCh38
rs1001599161 Chr.17: 18184316-18184316 on Build GRCh38
rs747813849 Chr.17: 18184358-18184363 on Build GRCh38
rs767011110 Chr.17: 18184439-18184439 on Build GRCh38
rs997201124 Chr.17: 18184253-18184253 on Build GRCh38
rs780747610 Chr.17: 18184357-18184365 on Build GRCh38
rs754349957 Chr.17: 18184297-18184297 on Build GRCh38
rs947253093 Chr.17: 18184360-18184361 on Build GRCh38
rs758262670 Chr.17: 18184342-18184343 on Build GRCh38
rs779379714 Chr.17: 18184367-18184375 on Build GRCh38
rs1029992487 Chr.17: 18184372-18184372 on Build GRCh38
rs776457581 Chr.17: 18184396-18184396 on Build GRCh38
rs913465156 Chr.17: 18184259-18184259 on Build GRCh38
rs377071193 Chr.17: 18184423-18184423 on Build GRCh38
rs963583867 Chr.17: 18184328-18184328 on Build GRCh38
rs900232835 Chr.17: 18184366-18184366 on Build GRCh38
rs1004309807 Chr.17: 18184331-18184348 on Build GRCh38
rs1032708221 Chr.17: 18184418-18184418 on Build GRCh38

View Full Product Details