Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 133450234-133450495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BFSP2
Entrez Gene ID
8419
Gene Name
beaded filament structural protein 2
Gene Aliases
CP47, CP49, CTRCT12, LIFL-L, PHAKOSIN
UniGene
Hs.659862
Gene Chromosome Location
Chr.3: 133397882-133475212 on Build GRCh38
Transcripts
NM_003571.3 XM_017007315.1 NR_135276.1 NR_135277.1 NR_135278.1

Gene Symbol
BFSP2-AS1
Entrez Gene ID
85003
Gene Name
BFSP2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.659202
Gene Chromosome Location
Chr.3: 133426402-133491146 on Build GRCh38
Transcripts
NM_003571.3 XM_017007315.1 NR_135276.1 NR_135277.1 NR_135278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4789553 c.867G>T p.E289D Chr.3: 133450440-133450440 on Build GRCh38
1184683 c.788C>T p.P263L Chr.3: 133450361-133450361 on Build GRCh38
1670582 c.848T>C p.V283A Chr.3: 133450421-133450421 on Build GRCh38
5355285 c.821C>T p.T274M Chr.3: 133450394-133450394 on Build GRCh38
5398188 c.830T>A p.I277N Chr.3: 133450403-133450403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749009723 Chr.3: 133450269-133450269 on Build GRCh38
rs992576618 Chr.3: 133450380-133450380 on Build GRCh38
rs751070406 Chr.3: 133450266-133450266 on Build GRCh38
rs199694156 Chr.3: 133450461-133450461 on Build GRCh38
rs770817384 Chr.3: 133450287-133450287 on Build GRCh38
rs760776920 Chr.3: 133450321-133450321 on Build GRCh38
rs541729511 Chr.3: 133450283-133450283 on Build GRCh38
rs745890103 Chr.3: 133450298-133450298 on Build GRCh38
rs148915741 Chr.3: 133450437-133450437 on Build GRCh38
rs1021803176 Chr.3: 133450413-133450413 on Build GRCh38
rs537111234 Chr.3: 133450273-133450273 on Build GRCh38
rs757058933 Chr.3: 133450418-133450418 on Build GRCh38
rs190627417 Chr.3: 133450434-133450434 on Build GRCh38
rs186574158 Chr.3: 133450394-133450394 on Build GRCh38
rs759028218 Chr.3: 133450372-133450372 on Build GRCh38
rs140944960 Chr.3: 133450304-133450304 on Build GRCh38
rs772818092 Chr.3: 133450268-133450269 on Build GRCh38
rs200703179 Chr.3: 133450334-133450334 on Build GRCh38
rs777516079 Chr.3: 133450395-133450395 on Build GRCh38
rs997318806 Chr.3: 133450263-133450263 on Build GRCh38
rs748970209 Chr.3: 133450398-133450398 on Build GRCh38
rs915912395 Chr.3: 133450456-133450456 on Build GRCh38
rs200862865 Chr.3: 133450440-133450440 on Build GRCh38
rs754268390 Chr.3: 133450259-133450259 on Build GRCh38
rs762266400 Chr.3: 133450358-133450358 on Build GRCh38
rs747132237 Chr.3: 133450433-133450433 on Build GRCh38
rs768718908 Chr.3: 133450435-133450435 on Build GRCh38
rs745801975 Chr.3: 133450425-133450425 on Build GRCh38
rs104893685 Chr.3: 133450432-133450432 on Build GRCh38
rs750976752 Chr.3: 133450369-133450369 on Build GRCh38
rs776810539 Chr.3: 133450353-133450353 on Build GRCh38
rs759768853 Chr.3: 133450290-133450292 on Build GRCh38
rs201642540 Chr.3: 133450384-133450384 on Build GRCh38
rs754536841 Chr.3: 133450267-133450267 on Build GRCh38
rs992864990 Chr.3: 133450445-133450445 on Build GRCh38
rs145444931 Chr.3: 133450438-133450438 on Build GRCh38
rs767108645 Chr.3: 133450379-133450379 on Build GRCh38
rs755834789 Chr.3: 133450270-133450270 on Build GRCh38
rs764424231 Chr.3: 133450329-133450329 on Build GRCh38
rs773755796 Chr.3: 133450453-133450453 on Build GRCh38
rs146257317 Chr.3: 133450420-133450420 on Build GRCh38
rs765858451 Chr.3: 133450262-133450262 on Build GRCh38
rs368762072 Chr.3: 133450268-133450268 on Build GRCh38
rs75746794 Chr.3: 133450296-133450296 on Build GRCh38
rs373196355 Chr.3: 133450279-133450279 on Build GRCh38
rs769687076 Chr.3: 133450263-133450263 on Build GRCh38
rs181275910 Chr.3: 133450323-133450323 on Build GRCh38
rs149735452 Chr.3: 133450363-133450363 on Build GRCh38

View Full Product Details