Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 68874716-68874973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN1
Entrez Gene ID
87
Gene Name
actinin alpha 1
Gene Aliases
BDPLT15
UniGene
Hs.235750
Gene Chromosome Location
Chr.14: 68874123-68979366 on Build GRCh38
Transcripts
NM_001102.3 NM_001130004.1 NM_001130005.1 XM_011537265.2 XM_011537266.2 XM_011537267.2 XM_011537268.2 XM_017021720.1 XM_017021721.1 XM_017021722.1 XM_017021723.1 XM_017021725.1 XM_017021726.1 XM_017021727.1 XM_017021728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7354088 c.2662G>A p.G888S Chr.14: 68874876-68874876 on Build GRCh38
4899190 c.2682C>T p.S894S Chr.14: 68874922-68874922 on Build GRCh38
4659128 c.2665G>A p.E889K Chr.14: 68874873-68874873 on Build GRCh38
4659130 c.2651C>T p.T884M Chr.14: 68874887-68874887 on Build GRCh38
6592966 c.2720C>T p.A907V Chr.14: 68874884-68874884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756160188 Chr.14: 68874929-68874929 on Build GRCh38
rs11557769 Chr.14: 68874936-68874936 on Build GRCh38
rs578254736 Chr.14: 68874736-68874736 on Build GRCh38
rs758102806 Chr.14: 68874900-68874900 on Build GRCh38
rs745859336 Chr.14: 68874923-68874923 on Build GRCh38
rs111795768 Chr.14: 68874862-68874862 on Build GRCh38
rs1013518228 Chr.14: 68874817-68874817 on Build GRCh38
rs541262717 Chr.14: 68874892-68874892 on Build GRCh38
rs774807508 Chr.14: 68874851-68874851 on Build GRCh38
rs758983643 Chr.14: 68874846-68874846 on Build GRCh38
rs545396188 Chr.14: 68874836-68874836 on Build GRCh38
rs1025278134 Chr.14: 68874819-68874819 on Build GRCh38
rs753420287 Chr.14: 68874810-68874810 on Build GRCh38
rs765483044 Chr.14: 68874802-68874802 on Build GRCh38
rs759103863 Chr.14: 68874877-68874877 on Build GRCh38
rs199901015 Chr.14: 68874953-68874953 on Build GRCh38
rs370350860 Chr.14: 68874840-68874840 on Build GRCh38
rs760633640 Chr.14: 68874786-68874786 on Build GRCh38
rs764889893 Chr.14: 68874884-68874884 on Build GRCh38
rs745956137 Chr.14: 68874839-68874839 on Build GRCh38
rs141668847 Chr.14: 68874904-68874904 on Build GRCh38
rs996505111 Chr.14: 68874820-68874820 on Build GRCh38
rs769389947 Chr.14: 68874944-68874944 on Build GRCh38
rs762175698 Chr.14: 68874854-68874854 on Build GRCh38
rs955084884 Chr.14: 68874853-68874853 on Build GRCh38
rs781108326 Chr.14: 68874921-68874921 on Build GRCh38
rs780144853 Chr.14: 68874932-68874932 on Build GRCh38
rs754618872 Chr.14: 68874824-68874824 on Build GRCh38
rs752368324 Chr.14: 68874899-68874899 on Build GRCh38
rs777665467 Chr.14: 68874828-68874828 on Build GRCh38
rs14582 Chr.14: 68874761-68874761 on Build GRCh38
rs1015876409 Chr.14: 68874847-68874847 on Build GRCh38
rs761305316 Chr.14: 68874943-68874943 on Build GRCh38
rs758246893 Chr.14: 68874826-68874826 on Build GRCh38
rs1033705565 Chr.14: 68874801-68874801 on Build GRCh38
rs192640536 Chr.14: 68874876-68874876 on Build GRCh38
rs547011909 Chr.14: 68874837-68874837 on Build GRCh38
rs773076904 Chr.14: 68874756-68874756 on Build GRCh38
rs773833507 Chr.14: 68874942-68874942 on Build GRCh38
rs757078973 Chr.14: 68874917-68874917 on Build GRCh38
rs746883304 Chr.14: 68874834-68874834 on Build GRCh38
rs1041886151 Chr.14: 68874744-68874744 on Build GRCh38
rs149327982 Chr.14: 68874827-68874827 on Build GRCh38
rs35738337 Chr.14: 68874737-68874738 on Build GRCh38
rs147023729 Chr.14: 68874933-68874933 on Build GRCh38
rs372486596 Chr.14: 68874825-68874825 on Build GRCh38
rs1000439473 Chr.14: 68874799-68874799 on Build GRCh38
rs757585091 Chr.14: 68874945-68874945 on Build GRCh38
rs763066535 Chr.14: 68874934-68874934 on Build GRCh38
rs77499007 Chr.14: 68874941-68874941 on Build GRCh38
rs781078800 Chr.14: 68874838-68874838 on Build GRCh38
rs768868163 Chr.14: 68874843-68874843 on Build GRCh38
rs763814462 Chr.14: 68874950-68874950 on Build GRCh38
rs749558617 Chr.14: 68874842-68874842 on Build GRCh38
rs962888664 Chr.14: 68874880-68874880 on Build GRCh38
rs375245539 Chr.14: 68874856-68874856 on Build GRCh38
rs903363952 Chr.14: 68874751-68874751 on Build GRCh38
rs1001606062 Chr.14: 68874737-68874737 on Build GRCh38

View Full Product Details