Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119894754-119895027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM30
Entrez Gene ID
11085
Gene Name
ADAM metallopeptidase domain 30
Gene Aliases
svph4
UniGene
Hs.283011
Gene Chromosome Location
Chr.1: 119893533-119896524 on Build GRCh38
Transcripts
NM_021794.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
894727 c.1480C>T p.R494C Chr.1: 119894857-119894857 on Build GRCh38
4659391 c.1574C>T p.A525V Chr.1: 119894763-119894763 on Build GRCh38
3862402 c.1474G>A p.E492K Chr.1: 119894863-119894863 on Build GRCh38
894726 c.1521C>T p.C507C Chr.1: 119894816-119894816 on Build GRCh38
1660406 c.1526G>A p.S509N Chr.1: 119894811-119894811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201440427 Chr.1: 119894791-119894791 on Build GRCh38
rs773033316 Chr.1: 119894783-119894783 on Build GRCh38
rs368574621 Chr.1: 119894923-119894923 on Build GRCh38
rs777017829 Chr.1: 119894985-119894985 on Build GRCh38
rs759535134 Chr.1: 119894801-119894801 on Build GRCh38
rs983558313 Chr.1: 119894874-119894874 on Build GRCh38
rs867694217 Chr.1: 119894922-119894922 on Build GRCh38
rs754320276 Chr.1: 119894837-119894837 on Build GRCh38
rs774183540 Chr.1: 119894882-119894882 on Build GRCh38
rs762875766 Chr.1: 119894785-119894785 on Build GRCh38
rs149279290 Chr.1: 119894857-119894857 on Build GRCh38
rs587649628 Chr.1: 119894816-119894816 on Build GRCh38
rs770650583 Chr.1: 119894961-119894961 on Build GRCh38
rs201129849 Chr.1: 119894848-119894848 on Build GRCh38
rs937654827 Chr.1: 119894986-119894986 on Build GRCh38
rs764425856 Chr.1: 119894916-119894916 on Build GRCh38
rs148460316 Chr.1: 119894929-119894929 on Build GRCh38
rs866185301 Chr.1: 119894950-119894950 on Build GRCh38
rs372067600 Chr.1: 119895000-119895000 on Build GRCh38
rs769611489 Chr.1: 119895005-119895016 on Build GRCh38
rs774429626 Chr.1: 119894787-119894787 on Build GRCh38
rs770832799 Chr.1: 119894871-119894871 on Build GRCh38
rs764520153 Chr.1: 119894817-119894817 on Build GRCh38
rs148305137 Chr.1: 119894856-119894856 on Build GRCh38
rs267597955 Chr.1: 119894884-119894884 on Build GRCh38
rs144613770 Chr.1: 119894981-119894981 on Build GRCh38
rs769559013 Chr.1: 119894782-119894782 on Build GRCh38
rs766359307 Chr.1: 119894786-119894786 on Build GRCh38
rs752884037 Chr.1: 119894813-119894813 on Build GRCh38
rs778815352 Chr.1: 119894964-119894964 on Build GRCh38
rs587690484 Chr.1: 119894853-119894853 on Build GRCh38
rs776647536 Chr.1: 119894992-119894992 on Build GRCh38
rs745759637 Chr.1: 119894971-119894971 on Build GRCh38
rs759610347 Chr.1: 119894885-119894885 on Build GRCh38
rs777403590 Chr.1: 119894931-119894931 on Build GRCh38
rs762302511 Chr.1: 119894921-119894921 on Build GRCh38
rs754189445 Chr.1: 119894917-119894917 on Build GRCh38
rs183935565 Chr.1: 119894926-119894926 on Build GRCh38
rs762145751 Chr.1: 119894991-119894991 on Build GRCh38
rs775697948 Chr.1: 119894972-119894972 on Build GRCh38
rs752265647 Chr.1: 119894927-119894927 on Build GRCh38
rs747851580 Chr.1: 119894843-119894844 on Build GRCh38
rs775753022 Chr.1: 119894896-119894896 on Build GRCh38
rs147643084 Chr.1: 119894980-119894980 on Build GRCh38
rs749091259 Chr.1: 119894867-119894867 on Build GRCh38
rs755701771 Chr.1: 119894930-119894930 on Build GRCh38
rs777527464 Chr.1: 119894858-119894858 on Build GRCh38
rs750875369 Chr.1: 119894924-119894924 on Build GRCh38
rs374608819 Chr.1: 119894905-119894905 on Build GRCh38
rs749039340 Chr.1: 119894941-119894941 on Build GRCh38

View Full Product Details