Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 140525374-140525639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMOD2
Entrez Gene ID
255520
Gene Name
ELMO domain containing 2
Gene Aliases
9830169G11Rik
UniGene
Hs.450105
Gene Chromosome Location
Chr.4: 140524146-140553770 on Build GRCh38
Transcripts
NM_153702.3 XM_005262885.3 XM_005262886.2 XM_011531818.2 XM_011531819.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4996397 c.142G>T p.E48* Chr.4: 140525570-140525570 on Build GRCh38
4122436 c.13T>G p.L5V Chr.4: 140525441-140525441 on Build GRCh38
1051452 c.124C>T p.Q42* Chr.4: 140525552-140525552 on Build GRCh38
480898 c.116T>C p.V39A Chr.4: 140525544-140525544 on Build GRCh38
3125749 c.140T>C p.I47T Chr.4: 140525568-140525568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775497995 Chr.4: 140525589-140525589 on Build GRCh38
rs141408415 Chr.4: 140525491-140525491 on Build GRCh38
rs781425851 Chr.4: 140525421-140525421 on Build GRCh38
rs755034557 Chr.4: 140525528-140525528 on Build GRCh38
rs760829336 Chr.4: 140525490-140525490 on Build GRCh38
rs201244894 Chr.4: 140525575-140525575 on Build GRCh38
rs35416675 Chr.4: 140525509-140525510 on Build GRCh38
rs769641417 Chr.4: 140525427-140525428 on Build GRCh38
rs773251277 Chr.4: 140525432-140525435 on Build GRCh38
rs746318253 Chr.4: 140525553-140525553 on Build GRCh38
rs762779518 Chr.4: 140525499-140525499 on Build GRCh38
rs752899297 Chr.4: 140525409-140525409 on Build GRCh38
rs780908581 Chr.4: 140525454-140525454 on Build GRCh38
rs771236943 Chr.4: 140525481-140525481 on Build GRCh38
rs551870214 Chr.4: 140525541-140525541 on Build GRCh38
rs370605685 Chr.4: 140525602-140525602 on Build GRCh38
rs773982149 Chr.4: 140525608-140525609 on Build GRCh38
rs541824231 Chr.4: 140525566-140525566 on Build GRCh38
rs749007094 Chr.4: 140525473-140525474 on Build GRCh38
rs912576639 Chr.4: 140525569-140525569 on Build GRCh38
rs752272494 Chr.4: 140525539-140525539 on Build GRCh38
rs541261073 Chr.4: 140525435-140525435 on Build GRCh38
rs750244517 Chr.4: 140525430-140525430 on Build GRCh38
rs3828535 Chr.4: 140525613-140525613 on Build GRCh38
rs1053923229 Chr.4: 140525473-140525473 on Build GRCh38
rs776891848 Chr.4: 140525604-140525604 on Build GRCh38
rs768090425 Chr.4: 140525582-140525582 on Build GRCh38
rs951112205 Chr.4: 140525410-140525410 on Build GRCh38
rs755049024 Chr.4: 140525404-140525405 on Build GRCh38
rs778227224 Chr.4: 140525479-140525479 on Build GRCh38
rs547216026 Chr.4: 140525548-140525548 on Build GRCh38
rs748081807 Chr.4: 140525509-140525509 on Build GRCh38
rs747121416 Chr.4: 140525595-140525595 on Build GRCh38
rs1041781372 Chr.4: 140525554-140525554 on Build GRCh38
rs750143144 Chr.4: 140525495-140525495 on Build GRCh38
rs780786101 Chr.4: 140525530-140525530 on Build GRCh38
rs764944668 Chr.4: 140525426-140525426 on Build GRCh38
rs1041797273 Chr.4: 140525431-140525431 on Build GRCh38
rs201983750 Chr.4: 140525457-140525457 on Build GRCh38
rs770780667 Chr.4: 140525494-140525494 on Build GRCh38
rs762001579 Chr.4: 140525612-140525612 on Build GRCh38
rs754489025 Chr.4: 140525449-140525449 on Build GRCh38
rs150918381 Chr.4: 140525526-140525526 on Build GRCh38
rs748077962 Chr.4: 140525420-140525421 on Build GRCh38
rs902907008 Chr.4: 140525447-140525447 on Build GRCh38
rs772580541 Chr.4: 140525565-140525565 on Build GRCh38
rs559119300 Chr.4: 140525448-140525448 on Build GRCh38
rs749689016 Chr.4: 140525480-140525480 on Build GRCh38
rs201497167 Chr.4: 140525489-140525489 on Build GRCh38
rs935760193 Chr.4: 140525472-140525472 on Build GRCh38
rs758218725 Chr.4: 140525432-140525432 on Build GRCh38
rs926535766 Chr.4: 140525496-140525496 on Build GRCh38
rs768764837 Chr.4: 140525599-140525599 on Build GRCh38
rs150922490 Chr.4: 140525544-140525544 on Build GRCh38
rs760015417 Chr.4: 140525488-140525488 on Build GRCh38
rs142469105 Chr.4: 140525542-140525542 on Build GRCh38
rs368666273 Chr.4: 140525414-140525414 on Build GRCh38
rs771311143 Chr.4: 140525547-140525547 on Build GRCh38
rs537745308 Chr.4: 140525523-140525523 on Build GRCh38

View Full Product Details