Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184766549-184766788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL1
Entrez Gene ID
2180
Gene Name
acyl-CoA synthetase long-chain family member 1
Gene Aliases
ACS1, FACL1, FACL2, LACS, LACS1, LACS2
UniGene
Hs.406678
Gene Chromosome Location
Chr.4: 184755595-184826593 on Build GRCh38
Transcripts
NM_001286708.1 NM_001286710.1 NM_001286711.1 NM_001286712.1 NM_001995.3 XM_005262828.1 XM_005262829.1 XM_005262831.1 XM_011531742.1 XM_017007887.1 XM_017007888.1 XM_017007889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
732933 c.1173C>A p.D391E Chr.4: 184766712-184766712 on Build GRCh38
3130329 c.1147A>T p.T383S Chr.4: 184766738-184766738 on Build GRCh38
3130328 c.1198G>A p.E400K Chr.4: 184766687-184766687 on Build GRCh38
1495707 c.1187G>T p.R396M Chr.4: 184766698-184766698 on Build GRCh38
3946226 c.1232T>A p.L411Q Chr.4: 184766653-184766653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376253989 Chr.4: 184766591-184766591 on Build GRCh38
rs11727009 Chr.4: 184766709-184766709 on Build GRCh38
rs779977377 Chr.4: 184766676-184766676 on Build GRCh38
rs763520587 Chr.4: 184766725-184766725 on Build GRCh38
rs755935233 Chr.4: 184766747-184766747 on Build GRCh38
rs763848224 Chr.4: 184766750-184766750 on Build GRCh38
rs767089434 Chr.4: 184766735-184766735 on Build GRCh38
rs58776258 Chr.4: 184766590-184766590 on Build GRCh38
rs753825034 Chr.4: 184766751-184766751 on Build GRCh38
rs749916942 Chr.4: 184766581-184766583 on Build GRCh38
rs765840018 Chr.4: 184766611-184766611 on Build GRCh38
rs748471468 Chr.4: 184766703-184766703 on Build GRCh38
rs776842655 Chr.4: 184766700-184766700 on Build GRCh38
rs538205726 Chr.4: 184766717-184766717 on Build GRCh38
rs766034538 Chr.4: 184766728-184766728 on Build GRCh38
rs745960555 Chr.4: 184766577-184766577 on Build GRCh38
rs144636771 Chr.4: 184766734-184766734 on Build GRCh38
rs749185265 Chr.4: 184766652-184766652 on Build GRCh38
rs760880493 Chr.4: 184766726-184766726 on Build GRCh38
rs768759701 Chr.4: 184766681-184766681 on Build GRCh38
rs751263695 Chr.4: 184766634-184766634 on Build GRCh38
rs762572035 Chr.4: 184766614-184766614 on Build GRCh38
rs752440254 Chr.4: 184766644-184766644 on Build GRCh38
rs772182808 Chr.4: 184766669-184766669 on Build GRCh38
rs750898273 Chr.4: 184766612-184766612 on Build GRCh38
rs781097855 Chr.4: 184766642-184766642 on Build GRCh38
rs371815814 Chr.4: 184766746-184766746 on Build GRCh38
rs776058596 Chr.4: 184766578-184766578 on Build GRCh38
rs755845769 Chr.4: 184766645-184766645 on Build GRCh38
rs769116781 Chr.4: 184766599-184766599 on Build GRCh38
rs762578199 Chr.4: 184766585-184766591 on Build GRCh38
rs776851663 Chr.4: 184766602-184766602 on Build GRCh38
rs559017153 Chr.4: 184766640-184766640 on Build GRCh38
rs773719585 Chr.4: 184766733-184766733 on Build GRCh38
rs116663524 Chr.4: 184766584-184766584 on Build GRCh38
rs142792812 Chr.4: 184766615-184766615 on Build GRCh38
rs762452670 Chr.4: 184766603-184766603 on Build GRCh38
rs575037653 Chr.4: 184766764-184766764 on Build GRCh38
rs372744784 Chr.4: 184766606-184766606 on Build GRCh38
rs775722898 Chr.4: 184766585-184766585 on Build GRCh38

View Full Product Details