Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 227530922-227531147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGFG1
Entrez Gene ID
3267
Gene Name
ArfGAP with FG repeats 1
Gene Aliases
HRB, RAB, RIP
UniGene
Hs.352962
Gene Chromosome Location
Chr.2: 227472169-227561222 on Build GRCh38
Transcripts
NM_001135187.1 NM_001135188.1 NM_001135189.1 NM_004504.4 XM_005246516.3 XM_006712478.3 XM_006712479.3 XM_006712480.3 XM_006712481.3 XM_017003954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3364618 c.709A>G p.N237D Chr.2: 227531105-227531105 on Build GRCh38
5886124 c.695-5T>C p.? Chr.2: 227531086-227531086 on Build GRCh38
5886125 c.695-5T>C p.? Chr.2: 227531086-227531086 on Build GRCh38
374399 c.714A>G p.A238A Chr.2: 227531110-227531110 on Build GRCh38
1591925 c.749G>T p.S250I Chr.2: 227531145-227531145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369535198 Chr.2: 227531053-227531053 on Build GRCh38
rs983150708 Chr.2: 227531019-227531019 on Build GRCh38
rs1052246423 Chr.2: 227530961-227530961 on Build GRCh38
rs781339447 Chr.2: 227531050-227531050 on Build GRCh38
rs567732797 Chr.2: 227531083-227531083 on Build GRCh38
rs996298868 Chr.2: 227531021-227531021 on Build GRCh38
rs913824314 Chr.2: 227530996-227530996 on Build GRCh38
rs772527880 Chr.2: 227531108-227531108 on Build GRCh38
rs760415448 Chr.2: 227531111-227531111 on Build GRCh38
rs533104610 Chr.2: 227530976-227530976 on Build GRCh38
rs1043097541 Chr.2: 227531009-227531009 on Build GRCh38
rs537105829 Chr.2: 227531015-227531015 on Build GRCh38
rs576614897 Chr.2: 227531030-227531030 on Build GRCh38
rs142891874 Chr.2: 227531033-227531033 on Build GRCh38
rs1014609088 Chr.2: 227531025-227531025 on Build GRCh38
rs565951137 Chr.2: 227530969-227530969 on Build GRCh38
rs749614328 Chr.2: 227531076-227531076 on Build GRCh38
rs551393957 Chr.2: 227530989-227530989 on Build GRCh38
rs569710903 Chr.2: 227531007-227531007 on Build GRCh38
rs1000791723 Chr.2: 227530944-227530944 on Build GRCh38
rs900344415 Chr.2: 227530995-227530995 on Build GRCh38
rs200923404 Chr.2: 227531081-227531081 on Build GRCh38
rs761286393 Chr.2: 227531088-227531088 on Build GRCh38
rs753713836 Chr.2: 227531118-227531118 on Build GRCh38
rs372044880 Chr.2: 227531066-227531066 on Build GRCh38
rs1050927190 Chr.2: 227531057-227531057 on Build GRCh38
rs780419811 Chr.2: 227531073-227531073 on Build GRCh38
rs757349205 Chr.2: 227531047-227531047 on Build GRCh38
rs1032565033 Chr.2: 227530975-227530975 on Build GRCh38
rs201909660 Chr.2: 227531105-227531105 on Build GRCh38
rs766077514 Chr.2: 227531112-227531112 on Build GRCh38
rs920059541 Chr.2: 227531035-227531035 on Build GRCh38
rs751644562 Chr.2: 227531046-227531046 on Build GRCh38

View Full Product Details