Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153469045-153469318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS7
Entrez Gene ID
55559
Gene Name
HAUS augmin like complex subunit 7
Gene Aliases
UCHL5IP, UIP1
UniGene
Hs.170835
Gene Chromosome Location
Chr.X: 153447665-153495525 on Build GRCh38
Transcripts
NM_017518.7 NR_073156.1

SNP Detail

SNP ID Chromosome Location
rs782587521 Chr.X: 153469272-153469272 on Build GRCh38
rs781873748 Chr.X: 153469236-153469236 on Build GRCh38
rs948519009 Chr.X: 153469247-153469247 on Build GRCh38
rs782056983 Chr.X: 153469175-153469175 on Build GRCh38
rs782188966 Chr.X: 153469116-153469116 on Build GRCh38
rs781886108 Chr.X: 153469104-153469104 on Build GRCh38
rs782336766 Chr.X: 153469117-153469117 on Build GRCh38
rs142766149 Chr.X: 153469126-153469126 on Build GRCh38
rs933569159 Chr.X: 153469096-153469096 on Build GRCh38
rs781834302 Chr.X: 153469292-153469292 on Build GRCh38
rs782777464 Chr.X: 153469230-153469230 on Build GRCh38
rs782543804 Chr.X: 153469106-153469106 on Build GRCh38
rs376652571 Chr.X: 153469228-153469228 on Build GRCh38
rs782328141 Chr.X: 153469177-153469177 on Build GRCh38
rs978344756 Chr.X: 153469109-153469109 on Build GRCh38
rs781788550 Chr.X: 153469249-153469249 on Build GRCh38
rs782148925 Chr.X: 153469240-153469240 on Build GRCh38
rs781956879 Chr.X: 153469226-153469226 on Build GRCh38
rs782604425 Chr.X: 153469121-153469121 on Build GRCh38
rs781843131 Chr.X: 153469099-153469099 on Build GRCh38
rs6643662 Chr.X: 153469291-153469291 on Build GRCh38
rs782044436 Chr.X: 153469130-153469130 on Build GRCh38
rs369890157 Chr.X: 153469241-153469241 on Build GRCh38
rs934362137 Chr.X: 153469135-153469135 on Build GRCh38
rs377655305 Chr.X: 153469090-153469090 on Build GRCh38
rs781905218 Chr.X: 153469086-153469086 on Build GRCh38
rs782464135 Chr.X: 153469251-153469251 on Build GRCh38
rs782575951 Chr.X: 153469111-153469111 on Build GRCh38
rs782411392 Chr.X: 153469127-153469127 on Build GRCh38

View Full Product Details