The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004283.3 | NP_004274.1 | 4 - 5 | 727 | 70 |
GenBank mRNA | AF081353.1 | - | 4 - 5 | 640 | 70 |
AF498933.1 | - | 3 - 4 | 466 | 70 | |
AK313538.1 | - | 4 - 5 | 727 | 70 | |
BC016471.1 | - | 4 - 5 | 704 | 70 |