Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
FHOD1 | NM_001318202.1 | 19 | 2739 | NP_001305131.1 |
NM_013241.2 | 17 | 2661 | NP_037373.2 | |
XM_006721180.1 | 8 | 1314 | XP_006721243.1 | |
XM_011523043.2 | 19 | 2955 | XP_011521345.1 | |
XM_011523044.1 | XP_011521346.1 | |||
XM_011523045.2 | 19 | 2449 | XP_011521347.1 | |
AB041046.1 | 17 | 2631 | BAD06250.1 | |
AB209584.1 | 15 | 3130 | BAD92821.1 | |
AF113615.1 | 17 | 2567 | AAD39906.1 | |
AK025992.1 | 3 | 351 | ||
AK301157.1 | 7 | 1372 | ||
AY192154.1 | 17 | 2549 | AAO38757.1 | |
BC033084.1 | 17 | 2606 | AAH33084.1 | |
KJ658417.1 | 19 | 2627 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr16:67153308 - 68071571 on Build GRCh38 | Gain | LOC100131303 KIAA0895L EXOC3L1 NUTF2 TMEM208 MIR328 SLC12A4 LRRC29 PLEKHG4 TRADD PSMB10 LOC107984813 THAP11 NRN1L NOL3 DUS2 ENKD1 LRRC36 C16orf86 LCAT CTRL SLC9A5 E2F4 GFOD2 DPEP2 FBXL8 PSKH1 CTCF HSF4 RANBP10 ZDHHC1 ELMO3 DPEP3 PARD6A EDC4 AGRP CENPT TSNAXIP1 DDX28 HSD11B2 LOC100505942 TPPP3 FAM65A ATP6V0D1 KCTD19 CARMIL2 ACD LOC105369155 FHOD1 |
nsv827707 | Chr16:67148355 - 67289296 on Build GRCh38 | Loss | KIAA0895L EXOC3L1 TMEM208 NOL3 MIR328 FBXL8 HSF4 ELMO3 LRRC29 SLC9A5 C16orf70 PLEKHG4 TRADD B3GNT9 E2F4 LOC105369155 FHOD1 |
nsv952044 | Chr16:67141198 - 67244397 on Build GRCh38 | Deletion | KIAA0895L EXOC3L1 TMEM208 NOL3 MIR328 FBXL8 HSF4 ELMO3 LRRC29 C16orf70 TRADD B3GNT9 E2F4 LOC105369155 FHOD1 |
nsv457513 | Chr16:67181417 - 67282697 on Build GRCh38 | Loss | KIAA0895L EXOC3L1 TMEM208 MIR328 PLEKHG4 E2F4 ELMO3 LOC105369155 FHOD1 LRRC29 SLC9A5 |
nsv524492 | Chr16:67174030 - 67294550 on Build GRCh38 | Loss | KIAA0895L EXOC3L1 TMEM208 NOL3 MIR328 ELMO3 LRRC29 SLC9A5 KCTD19 PLEKHG4 E2F4 LOC105369155 FHOD1 |
nsv524363 | Chr16:67136529 - 68071571 on Build GRCh38 | Loss | LOC100131303 KIAA0895L EXOC3L1 NUTF2 TMEM208 MIR328 SLC12A4 LRRC29 PLEKHG4 TRADD PSMB10 LOC107984813 THAP11 NRN1L NOL3 DUS2 ENKD1 LRRC36 C16orf86 LCAT CTRL SLC9A5 B3GNT9 E2F4 GFOD2 DPEP2 FBXL8 PSKH1 CTCF HSF4 RANBP10 ZDHHC1 ELMO3 DPEP3 PARD6A EDC4 AGRP CENPT TSNAXIP1 DDX28 HSD11B2 LOC100505942 TPPP3 FAM65A C16orf70 ATP6V0D1 KCTD19 CARMIL2 ACD LOC105369155 FHOD1 |