The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002276.4 | NP_002267.2 | 1 - 1 | 356 | 116 |
GenBank mRNA | AK313261.1 | - | 1 - 1 | 275 | 116 |
BC002539.2 | - | 1 - 1 | 260 | 116 | |
BC007628.1 | - | 1 - 1 | 260 | 116 | |
BC010409.2 | - | 1 - 1 | 267 | 116 | |
BC067744.1 | - | 1 - 1 | 275 | 116 | |
BC084574.1 | - | 1 - 1 | 258 | 116 | |
BG490639.1 | - | 1 - 1 | 260 | 116 | |
BG750971.1 | - | - | 349 | 116 | |
BP328445.1 | - | 1 - 1 | 116 | ||
DA068330.1 | - | 1 - 1 | 339 | 116 | |
Y00503.1 | - | 1 - 1 | 246 | 116 |