Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44888586-44888819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB2
Entrez Gene ID
3689
Gene Name
integrin subunit beta 2
Gene Aliases
CD18, LAD, LCAMB, LFA-1, MAC-1, MF17, MFI7
UniGene
Hs.375957
Gene Chromosome Location
Chr.21: 44885949-44928873 on Build GRCh38
Transcripts
NM_000211.4 NM_001127491.2 NM_001303238.1 XM_006724001.1 XM_017028341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7302055 c.2058C>G p.L686L Chr.21: 44888715-44888715 on Build GRCh38
350278 c.1975G>T p.G659C Chr.21: 44888798-44888798 on Build GRCh38
4917106 c.1990G>C p.E664Q Chr.21: 44888783-44888783 on Build GRCh38
1031368 c.2004G>A p.E668E Chr.21: 44888769-44888769 on Build GRCh38
1031366 c.2070T>C p.D690D Chr.21: 44888703-44888703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896459956 Chr.21: 44888717-44888717 on Build GRCh38
rs986681121 Chr.21: 44888746-44888746 on Build GRCh38
rs117884186 Chr.21: 44888715-44888715 on Build GRCh38
rs761724751 Chr.21: 44888793-44888793 on Build GRCh38
rs369283897 Chr.21: 44888748-44888748 on Build GRCh38
rs748923083 Chr.21: 44888778-44888778 on Build GRCh38
rs751290845 Chr.21: 44888654-44888654 on Build GRCh38
rs778278328 Chr.21: 44888749-44888749 on Build GRCh38
rs759270531 Chr.21: 44888645-44888645 on Build GRCh38
rs779520277 Chr.21: 44888674-44888674 on Build GRCh38
rs574393878 Chr.21: 44888635-44888635 on Build GRCh38
rs374293657 Chr.21: 44888744-44888744 on Build GRCh38
rs879017906 Chr.21: 44888700-44888700 on Build GRCh38
rs768143838 Chr.21: 44888781-44888781 on Build GRCh38
rs780361764 Chr.21: 44888722-44888722 on Build GRCh38
rs999711230 Chr.21: 44888706-44888706 on Build GRCh38
rs756282183 Chr.21: 44888721-44888721 on Build GRCh38
rs1044311307 Chr.21: 44888687-44888687 on Build GRCh38
rs376621955 Chr.21: 44888644-44888644 on Build GRCh38
rs147238010 Chr.21: 44888723-44888723 on Build GRCh38
rs748698954 Chr.21: 44888676-44888676 on Build GRCh38
rs764598463 Chr.21: 44888647-44888647 on Build GRCh38
rs772501506 Chr.21: 44888679-44888679 on Build GRCh38
rs573779039 Chr.21: 44888636-44888636 on Build GRCh38
rs565042973 Chr.21: 44888769-44888769 on Build GRCh38
rs762575142 Chr.21: 44888693-44888693 on Build GRCh38
rs1025458254 Chr.21: 44888738-44888738 on Build GRCh38
rs5844205 Chr.21: 44888694-44888694 on Build GRCh38
rs113221428 Chr.21: 44888692-44888692 on Build GRCh38
rs990892688 Chr.21: 44888617-44888617 on Build GRCh38
rs755049310 Chr.21: 44888727-44888727 on Build GRCh38
rs200241208 Chr.21: 44888664-44888664 on Build GRCh38
rs201448757 Chr.21: 44888684-44888684 on Build GRCh38
rs942375725 Chr.21: 44888760-44888760 on Build GRCh38
rs778865777 Chr.21: 44888732-44888732 on Build GRCh38
rs774832335 Chr.21: 44888689-44888689 on Build GRCh38
rs767506599 Chr.21: 44888798-44888798 on Build GRCh38
rs749447080 Chr.21: 44888671-44888671 on Build GRCh38
rs377664082 Chr.21: 44888651-44888651 on Build GRCh38
rs374429256 Chr.21: 44888650-44888650 on Build GRCh38
rs769010871 Chr.21: 44888672-44888672 on Build GRCh38
rs772955668 Chr.21: 44888801-44888801 on Build GRCh38
rs569637858 Chr.21: 44888616-44888616 on Build GRCh38
rs747419139 Chr.21: 44888767-44888767 on Build GRCh38
rs201725341 Chr.21: 44888668-44888668 on Build GRCh38
rs532445449 Chr.21: 44888790-44888790 on Build GRCh38
rs145972042 Chr.21: 44888772-44888772 on Build GRCh38
rs200335681 Chr.21: 44888695-44888695 on Build GRCh38
rs577771584 Chr.21: 44888660-44888660 on Build GRCh38
rs202213813 Chr.21: 44888669-44888669 on Build GRCh38
rs371539211 Chr.21: 44888733-44888733 on Build GRCh38
rs764134210 Chr.21: 44888652-44888652 on Build GRCh38
rs750578078 Chr.21: 44888665-44888665 on Build GRCh38
rs775160940 Chr.21: 44888777-44888777 on Build GRCh38
rs761741234 Chr.21: 44888709-44888709 on Build GRCh38
rs769558209 Chr.21: 44888685-44888685 on Build GRCh38
rs536911747 Chr.21: 44888628-44888628 on Build GRCh38

View Full Product Details