Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49451585-49451810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUT
Entrez Gene ID
4594
Gene Name
methylmalonyl-CoA mutase
Gene Aliases
MCM
UniGene
Hs.485527
Gene Chromosome Location
Chr.6: 49430360-49463328 on Build GRCh38
Transcripts
NM_000255.3 XM_005249143.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5915561 c.1087C>T p.P363S Chr.6: 49451711-49451711 on Build GRCh38
1080059 c.1208G>A p.R403Q Chr.6: 49451590-49451590 on Build GRCh38
2152418 c.1106G>A p.R369H Chr.6: 49451692-49451692 on Build GRCh38
484176 c.1192A>G p.T398A Chr.6: 49451606-49451606 on Build GRCh38
5472281 c.1107T>C p.R369R Chr.6: 49451691-49451691 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776065979 Chr.6: 49451705-49451705 on Build GRCh38
rs754742219 Chr.6: 49451662-49451662 on Build GRCh38
rs757863375 Chr.6: 49451616-49451616 on Build GRCh38
rs980824873 Chr.6: 49451754-49451754 on Build GRCh38
rs564069299 Chr.6: 49451692-49451692 on Build GRCh38
rs751208293 Chr.6: 49451729-49451729 on Build GRCh38
rs563776413 Chr.6: 49451707-49451707 on Build GRCh38
rs770946713 Chr.6: 49451684-49451684 on Build GRCh38
rs879253839 Chr.6: 49451716-49451716 on Build GRCh38
rs368790885 Chr.6: 49451690-49451690 on Build GRCh38
rs879253840 Chr.6: 49451634-49451634 on Build GRCh38
rs373027036 Chr.6: 49451726-49451726 on Build GRCh38
rs781020628 Chr.6: 49451664-49451664 on Build GRCh38
rs766010704 Chr.6: 49451636-49451636 on Build GRCh38
rs749884774 Chr.6: 49451614-49451614 on Build GRCh38
rs777583211 Chr.6: 49451682-49451682 on Build GRCh38
rs987439672 Chr.6: 49451755-49451755 on Build GRCh38
rs121918250 Chr.6: 49451668-49451668 on Build GRCh38
rs772552898 Chr.6: 49451693-49451693 on Build GRCh38
rs141825870 Chr.6: 49451764-49451764 on Build GRCh38
rs879253838 Chr.6: 49451715-49451715 on Build GRCh38
rs112089163 Chr.6: 49451783-49451783 on Build GRCh38
rs761129296 Chr.6: 49451706-49451706 on Build GRCh38
rs148091558 Chr.6: 49451673-49451673 on Build GRCh38
rs879253841 Chr.6: 49451616-49451617 on Build GRCh38
rs765852984 Chr.6: 49451728-49451728 on Build GRCh38
rs777031588 Chr.6: 49451724-49451724 on Build GRCh38
rs864309737 Chr.6: 49451701-49451701 on Build GRCh38
rs771021560 Chr.6: 49451649-49451650 on Build GRCh38
rs866933356 Chr.6: 49451641-49451641 on Build GRCh38
rs751206990 Chr.6: 49451659-49451659 on Build GRCh38
rs755977989 Chr.6: 49451679-49451679 on Build GRCh38
rs767983689 Chr.6: 49451708-49451708 on Build GRCh38
rs150968643 Chr.6: 49451683-49451683 on Build GRCh38
rs376009062 Chr.6: 49451757-49451757 on Build GRCh38
rs754689525 Chr.6: 49451735-49451735 on Build GRCh38

View Full Product Details