Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74919358-74919585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USH1G
Entrez Gene ID
124590
Gene Name
USH1 protein network component sans
Gene Aliases
ANKS4A, SANS
UniGene
Hs.376688
Gene Chromosome Location
Chr.17: 74916083-74923263 on Build GRCh38
Transcripts
NM_001282489.2 NM_173477.4 XM_011524296.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5456645 c.1361C>T p.P454L Chr.17: 74919475-74919475 on Build GRCh38
2978200 c.1280G>A p.R427H Chr.17: 74919556-74919556 on Build GRCh38
4525401 c.1329G>A p.V443V Chr.17: 74919507-74919507 on Build GRCh38
5003848 c.1291G>T p.V431F Chr.17: 74919545-74919545 on Build GRCh38
1385864 c.1283G>A p.S428N Chr.17: 74919553-74919553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765736586 Chr.17: 74919431-74919431 on Build GRCh38
rs551386496 Chr.17: 74919447-74919447 on Build GRCh38
rs777486419 Chr.17: 74919539-74919539 on Build GRCh38
rs971548417 Chr.17: 74919490-74919490 on Build GRCh38
rs989804696 Chr.17: 74919523-74919523 on Build GRCh38
rs114720069 Chr.17: 74919385-74919385 on Build GRCh38
rs747285177 Chr.17: 74919475-74919475 on Build GRCh38
rs943263343 Chr.17: 74919555-74919555 on Build GRCh38
rs763508231 Chr.17: 74919499-74919499 on Build GRCh38
rs765888502 Chr.17: 74919417-74919417 on Build GRCh38
rs760743915 Chr.17: 74919409-74919409 on Build GRCh38
rs753794521 Chr.17: 74919545-74919545 on Build GRCh38
rs970523771 Chr.17: 74919437-74919437 on Build GRCh38
rs375297515 Chr.17: 74919451-74919451 on Build GRCh38
rs757101131 Chr.17: 74919550-74919550 on Build GRCh38
rs752513148 Chr.17: 74919493-74919494 on Build GRCh38
rs757991416 Chr.17: 74919423-74919423 on Build GRCh38
rs1004192113 Chr.17: 74919432-74919432 on Build GRCh38
rs755838297 Chr.17: 74919500-74919501 on Build GRCh38
rs776814552 Chr.17: 74919411-74919411 on Build GRCh38
rs751073735 Chr.17: 74919510-74919510 on Build GRCh38
rs759065977 Chr.17: 74919518-74919518 on Build GRCh38
rs201359669 Chr.17: 74919496-74919496 on Build GRCh38
rs762249466 Chr.17: 74919416-74919416 on Build GRCh38
rs886043626 Chr.17: 74919525-74919525 on Build GRCh38
rs775795179 Chr.17: 74919404-74919404 on Build GRCh38
rs778876458 Chr.17: 74919551-74919551 on Build GRCh38
rs773872479 Chr.17: 74919497-74919497 on Build GRCh38
rs754536324 Chr.17: 74919435-74919435 on Build GRCh38
rs373882524 Chr.17: 74919434-74919434 on Build GRCh38
rs752343151 Chr.17: 74919529-74919529 on Build GRCh38
rs776968656 Chr.17: 74919481-74919481 on Build GRCh38
rs991658805 Chr.17: 74919449-74919449 on Build GRCh38
rs368110384 Chr.17: 74919557-74919557 on Build GRCh38
rs775382564 Chr.17: 74919465-74919465 on Build GRCh38
rs368389700 Chr.17: 74919436-74919436 on Build GRCh38
rs780954300 Chr.17: 74919428-74919429 on Build GRCh38
rs748326974 Chr.17: 74919564-74919564 on Build GRCh38
rs750965243 Chr.17: 74919420-74919420 on Build GRCh38
rs765949002 Chr.17: 74919500-74919500 on Build GRCh38
rs762322942 Chr.17: 74919482-74919482 on Build GRCh38
rs914830331 Chr.17: 74919503-74919503 on Build GRCh38
rs755890365 Chr.17: 74919438-74919438 on Build GRCh38
rs768787249 Chr.17: 74919401-74919401 on Build GRCh38
rs955918806 Chr.17: 74919522-74919522 on Build GRCh38
rs764553015 Chr.17: 74919410-74919410 on Build GRCh38
rs397517925 Chr.17: 74919463-74919463 on Build GRCh38
rs372580540 Chr.17: 74919478-74919478 on Build GRCh38
rs767134204 Chr.17: 74919519-74919519 on Build GRCh38
rs924339909 Chr.17: 74919505-74919505 on Build GRCh38
rs962741334 Chr.17: 74919435-74919436 on Build GRCh38
rs781625717 Chr.17: 74919563-74919563 on Build GRCh38
rs755908331 Chr.17: 74919530-74919530 on Build GRCh38

View Full Product Details