The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323281.1 | NP_001310210.1 | 1 - 2 | 443 | 75 |
NM_001323282.1 | NP_001310211.1 | 1 - 2 | 443 | 75 | |
NM_002895.4 | NP_002886.2 | 1 - 2 | 443 | 75 | |
NM_183404.3 | NP_899662.1 | 1 - 2 | 443 | 75 | |
XM_006723842.2 | XP_006723905.1 | 1 - 2 | 219 | 75 | |
XM_011528955.2 | XP_011527257.1 | 1 - 2 | 219 | 75 | |
XM_011528958.2 | XP_011527260.1 | 1 - 2 | 219 | 75 | |
XM_017027992.1 | XP_016883481.1 | 1 - 2 | 257 | 75 | |
GenBank mRNA | AK290380.1 | - | 1 - 2 | 219 | 75 |
AK308096.1 | - | 1 - 2 | 203 | 75 | |
AK310870.1 | - | 1 - 2 | 228 | 75 | |
BC032247.1 | - | 1 - 2 | 226 | 75 | |
DC375054.1 | - | 1 - 2 | 445 | 75 | |
L14812.1 | - | 1 - 2 | 224 | 75 |