Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2U1
Entrez Gene ID
113612
Gene Name
cytochrome P450 family 2 subfamily U member 1
Gene Aliases
P450TEC, SPG49, SPG56
Chromosome Location
Chr.4: 107931549 - 107953461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183075.2 NP_898898.1 1 - 2 574 63
GenBank mRNA AK290002.1 - 1 - 2 579 63
AK303284.1 - 2 - 3 379 63
AK304724.1 - 2 - 3 452 63
AY343323.1 - 1 - 2 530 63
BC132767.1 - 1 - 2 539 63
BC136483.1 - 1 - 2 539 63
BX354124.2 - 1 - 2 511 63
BX437421.2 - 1 - 2 765 63

View Full Product Details