Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144512225-144512478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RECQL4
Entrez Gene ID
9401
Gene Name
RecQ like helicase 4
Gene Aliases
RECQ4
UniGene
Hs.31442
Gene Chromosome Location
Chr.8: 144511284-144517833 on Build GRCh38
Transcripts
NM_004260.3 XM_011517384.2 XM_017013991.1 XM_017013992.1 XM_017013993.1 XM_017013994.1 XM_017013995.1 XM_017013996.1 XM_017013997.1 XM_017013998.1 XM_017013999.1 XM_017014000.1 XM_017014001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2871877 c.2987T>C p.M996T Chr.8: 144512460-144512460 on Build GRCh38
1313870 c.3136G>A p.E1046K Chr.8: 144512244-144512244 on Build GRCh38
6764678 c.3055G>T p.G1019C Chr.8: 144512392-144512392 on Build GRCh38
6963891 c.3011G>A p.R1004Q Chr.8: 144512436-144512436 on Build GRCh38
6764680 c.3061C>T p.R1021W Chr.8: 144512319-144512319 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008463415 Chr.8: 144512376-144512376 on Build GRCh38
rs767786765 Chr.8: 144512293-144512293 on Build GRCh38
rs372511135 Chr.8: 144512385-144512385 on Build GRCh38
rs754479836 Chr.8: 144512308-144512308 on Build GRCh38
rs761975311 Chr.8: 144512280-144512280 on Build GRCh38
rs952414985 Chr.8: 144512297-144512297 on Build GRCh38
rs779017449 Chr.8: 144512386-144512386 on Build GRCh38
rs398124118 Chr.8: 144512252-144512252 on Build GRCh38
rs373739637 Chr.8: 144512307-144512307 on Build GRCh38
rs767876394 Chr.8: 144512290-144512290 on Build GRCh38
rs773619560 Chr.8: 144512334-144512334 on Build GRCh38
rs369172759 Chr.8: 144512449-144512449 on Build GRCh38
rs758067843 Chr.8: 144512364-144512364 on Build GRCh38
rs368585080 Chr.8: 144512391-144512391 on Build GRCh38
rs903820228 Chr.8: 144512349-144512349 on Build GRCh38
rs377031190 Chr.8: 144512387-144512387 on Build GRCh38
rs758008013 Chr.8: 144512316-144512316 on Build GRCh38
rs373238823 Chr.8: 144512248-144512248 on Build GRCh38
rs760096494 Chr.8: 144512407-144512407 on Build GRCh38
rs758339361 Chr.8: 144512447-144512447 on Build GRCh38
rs940837869 Chr.8: 144512317-144512317 on Build GRCh38
rs374254271 Chr.8: 144512267-144512267 on Build GRCh38
rs370954099 Chr.8: 144512304-144512304 on Build GRCh38
rs759912361 Chr.8: 144512292-144512292 on Build GRCh38
rs753381594 Chr.8: 144512305-144512305 on Build GRCh38
rs772220332 Chr.8: 144512333-144512333 on Build GRCh38
rs35348691 Chr.8: 144512247-144512247 on Build GRCh38
rs771832340 Chr.8: 144512260-144512260 on Build GRCh38
rs767281689 Chr.8: 144512435-144512435 on Build GRCh38
rs778762414 Chr.8: 144512249-144512249 on Build GRCh38
rs368982201 Chr.8: 144512434-144512434 on Build GRCh38
rs762312510 Chr.8: 144512425-144512425 on Build GRCh38
rs114149451 Chr.8: 144512322-144512322 on Build GRCh38
rs137853232 Chr.8: 144512319-144512319 on Build GRCh38
rs748382510 Chr.8: 144512330-144512330 on Build GRCh38
rs768685242 Chr.8: 144512325-144512325 on Build GRCh38
rs898859819 Chr.8: 144512409-144512409 on Build GRCh38
rs750261857 Chr.8: 144512436-144512436 on Build GRCh38
rs4925828 Chr.8: 144512253-144512253 on Build GRCh38
rs778030723 Chr.8: 144512448-144512448 on Build GRCh38
rs902652277 Chr.8: 144512450-144512450 on Build GRCh38
rs764754383 Chr.8: 144512358-144512358 on Build GRCh38
rs36023964 Chr.8: 144512437-144512437 on Build GRCh38
rs750400159 Chr.8: 144512405-144512405 on Build GRCh38
rs771381171 Chr.8: 144512404-144512404 on Build GRCh38
rs760150078 Chr.8: 144512357-144512357 on Build GRCh38
rs999814906 Chr.8: 144512456-144512456 on Build GRCh38
rs786200889 Chr.8: 144512326-144512326 on Build GRCh38
rs948348185 Chr.8: 144512338-144512338 on Build GRCh38
rs745759208 Chr.8: 144512379-144512379 on Build GRCh38
rs4251691 Chr.8: 144512433-144512433 on Build GRCh38
rs770897193 Chr.8: 144512266-144512266 on Build GRCh38
rs767057306 Chr.8: 144512301-144512301 on Build GRCh38
rs774570166 Chr.8: 144512278-144512278 on Build GRCh38
rs756983251 Chr.8: 144512372-144512372 on Build GRCh38
rs763712794 Chr.8: 144512367-144512367 on Build GRCh38
rs182454890 Chr.8: 144512368-144512368 on Build GRCh38
rs372660887 Chr.8: 144512420-144512420 on Build GRCh38
rs768740437 Chr.8: 144512277-144512277 on Build GRCh38
rs772320981 Chr.8: 144512388-144512388 on Build GRCh38
rs34919378 Chr.8: 144512438-144512438 on Build GRCh38
rs768374433 Chr.8: 144512320-144512320 on Build GRCh38
rs747239208 Chr.8: 144512398-144512398 on Build GRCh38
rs374983038 Chr.8: 144512335-144512335 on Build GRCh38
rs777211987 Chr.8: 144512346-144512346 on Build GRCh38
rs918047641 Chr.8: 144512445-144512445 on Build GRCh38
rs774320251 Chr.8: 144512329-144512329 on Build GRCh38
rs560476442 Chr.8: 144512315-144512315 on Build GRCh38
rs763665817 Chr.8: 144512431-144512431 on Build GRCh38
rs376504280 Chr.8: 144512291-144512291 on Build GRCh38
rs766859426 Chr.8: 144512343-144512343 on Build GRCh38
rs34666647 Chr.8: 144512318-144512318 on Build GRCh38
rs771538008 Chr.8: 144512307-144512308 on Build GRCh38
rs370867731 Chr.8: 144512261-144512261 on Build GRCh38
rs948234493 Chr.8: 144512458-144512458 on Build GRCh38
rs765732606 Chr.8: 144512415-144512415 on Build GRCh38
rs386833852 Chr.8: 144512308-144512308 on Build GRCh38
rs764657835 Chr.8: 144512312-144512312 on Build GRCh38
rs781000093 Chr.8: 144512375-144512375 on Build GRCh38
rs752195344 Chr.8: 144512359-144512359 on Build GRCh38
rs763078132 Chr.8: 144512268-144512268 on Build GRCh38

View Full Product Details