The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027991.1 | 1 - 2 | 1665 | 142 | |
GenBank mRNA | AK074514.1 | - | 2 - 3 | 1107 | 142 |
BC032688.1 | - | 1 - 2 | 1447 | 142 | |
X89271.1 | - | 1 - 2 | 1525 | 142 |