Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM38
Entrez Gene ID
55544
Gene Name
RNA binding motif protein 38
Gene Aliases
HSRNASEB, RNPC1, SEB4B, SEB4D, dJ800J21.2
Chromosome Location
Chr.20: 57391398 - 57409333 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291780.1 NP_001278709.1 4 - 5 695 140
NM_017495.5 NP_059965.2 3 - 4 599 140
GenBank mRNA AF432218.1 - 3 - 4 595 140
BC018711.1 - 3 - 4 351 140
BE840212.1 - 1 - 2 140
BF371746.1 - 1 - 2 140
BM549757.1 - - 594 140
X75314.1 - 3 - 4 386 140

View Full Product Details