Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50953802-50954075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
UniGene
Hs.306291
Gene Chromosome Location
Chr.13: 50909678-50970462 on Build GRCh38
Transcripts
NM_001142279.2 NM_024570.3 XM_006719867.3 XM_011535229.1 XM_011535233.2 XM_017020747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6404930 c.761A>G p.E254G Chr.13: 50953924-50953924 on Build GRCh38
7325110 c.770A>G p.E257G Chr.13: 50953933-50953933 on Build GRCh38
1300243 c.787A>G p.T263A Chr.13: 50953950-50953950 on Build GRCh38
328852 c.777A>T p.K259N Chr.13: 50953940-50953940 on Build GRCh38
6139073 c.789T>C p.T263T Chr.13: 50953952-50953952 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs146378238 Chr.13: 50953925-50953925 on Build GRCh38
rs761333168 Chr.13: 50953990-50953990 on Build GRCh38
rs756850979 Chr.13: 50953887-50953887 on Build GRCh38
rs771299667 Chr.13: 50953929-50953929 on Build GRCh38
rs189116235 Chr.13: 50954029-50954029 on Build GRCh38
rs866929388 Chr.13: 50953847-50953847 on Build GRCh38
rs749536701 Chr.13: 50953868-50953868 on Build GRCh38
rs775119865 Chr.13: 50953971-50953971 on Build GRCh38
rs755138594 Chr.13: 50953885-50953885 on Build GRCh38
rs369199159 Chr.13: 50954015-50954015 on Build GRCh38
rs748144224 Chr.13: 50953907-50953907 on Build GRCh38
rs778980232 Chr.13: 50954014-50954014 on Build GRCh38
rs757200421 Chr.13: 50953993-50953993 on Build GRCh38
rs150363383 Chr.13: 50953950-50953950 on Build GRCh38
rs767399621 Chr.13: 50954000-50954000 on Build GRCh38
rs747575443 Chr.13: 50954023-50954023 on Build GRCh38
rs779113809 Chr.13: 50953893-50953893 on Build GRCh38
rs755266257 Chr.13: 50954007-50954007 on Build GRCh38
rs1043966687 Chr.13: 50953999-50953999 on Build GRCh38
rs972687891 Chr.13: 50953936-50953936 on Build GRCh38
rs777772670 Chr.13: 50954022-50954022 on Build GRCh38
rs183734174 Chr.13: 50953951-50953951 on Build GRCh38
rs763654606 Chr.13: 50953981-50953981 on Build GRCh38
rs776847998 Chr.13: 50953949-50953949 on Build GRCh38
rs780778643 Chr.13: 50953865-50953865 on Build GRCh38
rs750179605 Chr.13: 50954001-50954001 on Build GRCh38
rs747450548 Chr.13: 50953926-50953926 on Build GRCh38
rs371349565 Chr.13: 50953867-50953867 on Build GRCh38
rs757145306 Chr.13: 50953858-50953858 on Build GRCh38
rs76413207 Chr.13: 50953991-50953991 on Build GRCh38
rs191265144 Chr.13: 50954030-50954030 on Build GRCh38
rs113879083 Chr.13: 50953835-50953836 on Build GRCh38
rs113827426 Chr.13: 50954035-50954035 on Build GRCh38
rs200721645 Chr.13: 50953919-50953919 on Build GRCh38

View Full Product Details