Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38349241-38349488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRED1
Entrez Gene ID
161742
Gene Name
sprouty related EVH1 domain containing 1
Gene Aliases
NFLS, PPP1R147, hSpred1, spred-1
UniGene
Hs.525781
Gene Chromosome Location
Chr.15: 38252087-38357249 on Build GRCh38
Transcripts
NM_152594.2 XM_005254202.3 XM_011521289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
238234 c.633C>T p.Y211Y Chr.15: 38349472-38349472 on Build GRCh38
6789281 c.641G>A p.R214Q Chr.15: 38349480-38349480 on Build GRCh38
4054220 c.634G>A p.V212I Chr.15: 38349473-38349473 on Build GRCh38
5036928 c.618C>T p.S206S Chr.15: 38349457-38349457 on Build GRCh38
1707997 c.613C>T p.Q205* Chr.15: 38349452-38349452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368660900 Chr.15: 38349458-38349458 on Build GRCh38
rs535098540 Chr.15: 38349281-38349281 on Build GRCh38
rs902023448 Chr.15: 38349289-38349289 on Build GRCh38
rs371902444 Chr.15: 38349407-38349407 on Build GRCh38
rs772936179 Chr.15: 38349389-38349389 on Build GRCh38
rs1029233073 Chr.15: 38349358-38349358 on Build GRCh38
rs952232537 Chr.15: 38349355-38349355 on Build GRCh38
rs996297376 Chr.15: 38349292-38349292 on Build GRCh38
rs370015720 Chr.15: 38349385-38349385 on Build GRCh38
rs761697388 Chr.15: 38349380-38349380 on Build GRCh38
rs751598800 Chr.15: 38349410-38349410 on Build GRCh38
rs115970207 Chr.15: 38349415-38349415 on Build GRCh38
rs763102882 Chr.15: 38349406-38349406 on Build GRCh38
rs746708059 Chr.15: 38349457-38349457 on Build GRCh38
rs183730237 Chr.15: 38349364-38349364 on Build GRCh38
rs755068474 Chr.15: 38349414-38349414 on Build GRCh38
rs779879921 Chr.15: 38349453-38349453 on Build GRCh38
rs147474792 Chr.15: 38349426-38349426 on Build GRCh38
rs756643495 Chr.15: 38349433-38349433 on Build GRCh38
rs993615089 Chr.15: 38349331-38349331 on Build GRCh38
rs778144652 Chr.15: 38349438-38349438 on Build GRCh38
rs758087535 Chr.15: 38349449-38349449 on Build GRCh38
rs139897054 Chr.15: 38349446-38349446 on Build GRCh38
rs1002159507 Chr.15: 38349273-38349273 on Build GRCh38
rs896528625 Chr.15: 38349309-38349309 on Build GRCh38
rs1026869903 Chr.15: 38349343-38349343 on Build GRCh38
rs553337721 Chr.15: 38349361-38349361 on Build GRCh38
rs776073446 Chr.15: 38349463-38349463 on Build GRCh38

View Full Product Details