Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184189261-184189528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCF3
Entrez Gene ID
55324
Gene Name
ATP binding cassette subfamily F member 3
Gene Aliases
EST201864
UniGene
Hs.361323
Gene Chromosome Location
Chr.3: 184186075-184194012 on Build GRCh38
Transcripts
NM_018358.2 XM_011512959.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6590666 c.1110G>A p.L370L Chr.3: 184189440-184189440 on Build GRCh38
5974667 c.1057G>A p.E353K Chr.3: 184189277-184189277 on Build GRCh38
278552 c.1100A>G p.E367G Chr.3: 184189430-184189430 on Build GRCh38
729575 c.1045C>T p.L349L Chr.3: 184189265-184189265 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771057467 Chr.3: 184189337-184189337 on Build GRCh38
rs371454488 Chr.3: 184189505-184189505 on Build GRCh38
rs376539324 Chr.3: 184189380-184189380 on Build GRCh38
rs184083437 Chr.3: 184189368-184189368 on Build GRCh38
rs369013869 Chr.3: 184189365-184189365 on Build GRCh38
rs754809586 Chr.3: 184189433-184189433 on Build GRCh38
rs1006601439 Chr.3: 184189411-184189411 on Build GRCh38
rs191513871 Chr.3: 184189319-184189319 on Build GRCh38
rs201777346 Chr.3: 184189483-184189483 on Build GRCh38
rs750053812 Chr.3: 184189410-184189410 on Build GRCh38
rs1030574511 Chr.3: 184189333-184189333 on Build GRCh38
rs763415144 Chr.3: 184189495-184189495 on Build GRCh38
rs761159122 Chr.3: 184189377-184189377 on Build GRCh38
rs919264090 Chr.3: 184189499-184189499 on Build GRCh38
rs757246284 Chr.3: 184189459-184189459 on Build GRCh38
rs746122230 Chr.3: 184189364-184189364 on Build GRCh38
rs745995212 Chr.3: 184189464-184189464 on Build GRCh38
rs766194967 Chr.3: 184189417-184189417 on Build GRCh38
rs777083765 Chr.3: 184189466-184189467 on Build GRCh38
rs781261708 Chr.3: 184189315-184189315 on Build GRCh38
rs202133471 Chr.3: 184189437-184189437 on Build GRCh38
rs992071661 Chr.3: 184189503-184189503 on Build GRCh38
rs764823398 Chr.3: 184189394-184189394 on Build GRCh38
rs780933877 Chr.3: 184189438-184189438 on Build GRCh38
rs200262122 Chr.3: 184189430-184189430 on Build GRCh38
rs772266728 Chr.3: 184189471-184189471 on Build GRCh38
rs769037857 Chr.3: 184189379-184189379 on Build GRCh38
rs779059609 Chr.3: 184189346-184189346 on Build GRCh38
rs552347254 Chr.3: 184189382-184189382 on Build GRCh38
rs769074174 Chr.3: 184189490-184189490 on Build GRCh38
rs371757934 Chr.3: 184189297-184189297 on Build GRCh38
rs752702570 Chr.3: 184189451-184189451 on Build GRCh38
rs754861764 Chr.3: 184189306-184189306 on Build GRCh38
rs951245702 Chr.3: 184189502-184189502 on Build GRCh38
rs762676043 Chr.3: 184189415-184189415 on Build GRCh38
rs113643335 Chr.3: 184189370-184189370 on Build GRCh38
rs780321460 Chr.3: 184189482-184189482 on Build GRCh38

View Full Product Details