Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41061644-41061901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC2
Entrez Gene ID
10930
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 2
Gene Aliases
ARCD1, ARP1
UniGene
Hs.555915
Gene Chromosome Location
Chr.6: 41053201-41064891 on Build GRCh38
Transcripts
NM_006789.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3078322 c.464T>C p.F155S Chr.6: 41061660-41061660 on Build GRCh38
5040802 c.477G>C p.E159D Chr.6: 41061673-41061673 on Build GRCh38
741949 c.577G>T p.E193* Chr.6: 41061773-41061773 on Build GRCh38
4662505 c.646G>A p.E216K Chr.6: 41061842-41061842 on Build GRCh38
7236379 c.673T>C p.*225Q Chr.6: 41061869-41061869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976731769 Chr.6: 41061785-41061785 on Build GRCh38
rs771082334 Chr.6: 41061686-41061686 on Build GRCh38
rs749854072 Chr.6: 41061726-41061726 on Build GRCh38
rs1018706795 Chr.6: 41061727-41061727 on Build GRCh38
rs1040047097 Chr.6: 41061670-41061670 on Build GRCh38
rs1065197 Chr.6: 41061749-41061749 on Build GRCh38
rs754439371 Chr.6: 41061838-41061838 on Build GRCh38
rs534595473 Chr.6: 41061748-41061748 on Build GRCh38
rs757783760 Chr.6: 41061862-41061862 on Build GRCh38
rs922722488 Chr.6: 41061786-41061786 on Build GRCh38
rs749493226 Chr.6: 41061782-41061782 on Build GRCh38
rs748139491 Chr.6: 41061794-41061794 on Build GRCh38
rs745865989 Chr.6: 41061763-41061763 on Build GRCh38
rs769941939 Chr.6: 41061705-41061705 on Build GRCh38
rs768429186 Chr.6: 41061717-41061717 on Build GRCh38
rs761629533 Chr.6: 41061725-41061725 on Build GRCh38
rs900245360 Chr.6: 41061682-41061682 on Build GRCh38
rs746120000 Chr.6: 41061701-41061701 on Build GRCh38
rs140342245 Chr.6: 41061676-41061676 on Build GRCh38
rs780079400 Chr.6: 41061874-41061874 on Build GRCh38
rs193261337 Chr.6: 41061723-41061723 on Build GRCh38
rs758980965 Chr.6: 41061671-41061671 on Build GRCh38
rs768484237 Chr.6: 41061784-41061784 on Build GRCh38
rs41273364 Chr.6: 41061871-41061871 on Build GRCh38
rs374264492 Chr.6: 41061839-41061839 on Build GRCh38
rs781342303 Chr.6: 41061762-41061762 on Build GRCh38
rs762799569 Chr.6: 41061716-41061716 on Build GRCh38
rs114127156 Chr.6: 41061791-41061791 on Build GRCh38
rs780315538 Chr.6: 41061774-41061774 on Build GRCh38
rs371329415 Chr.6: 41061707-41061707 on Build GRCh38
rs760417585 Chr.6: 41061730-41061730 on Build GRCh38
rs776461790 Chr.6: 41061841-41061841 on Build GRCh38
rs1001142508 Chr.6: 41061669-41061669 on Build GRCh38
rs775275042 Chr.6: 41061842-41061844 on Build GRCh38
rs776505763 Chr.6: 41061742-41061742 on Build GRCh38
rs758881992 Chr.6: 41061742-41061742 on Build GRCh38
rs988316925 Chr.6: 41061810-41061810 on Build GRCh38
rs150558136 Chr.6: 41061842-41061842 on Build GRCh38
rs769856886 Chr.6: 41061767-41061767 on Build GRCh38
rs760329698 Chr.6: 41061833-41061833 on Build GRCh38
rs62396282 Chr.6: 41061759-41061759 on Build GRCh38
rs543353890 Chr.6: 41061868-41061868 on Build GRCh38
rs368093639 Chr.6: 41061851-41061851 on Build GRCh38
rs892960734 Chr.6: 41061752-41061752 on Build GRCh38
rs757652639 Chr.6: 41061754-41061754 on Build GRCh38
rs767972984 Chr.6: 41061865-41061865 on Build GRCh38
rs993993833 Chr.6: 41061688-41061688 on Build GRCh38
rs138132132 Chr.6: 41061675-41061675 on Build GRCh38
rs753055205 Chr.6: 41061667-41061667 on Build GRCh38
rs367590589 Chr.6: 41061695-41061695 on Build GRCh38
rs772784527 Chr.6: 41061827-41061827 on Build GRCh38
rs766065060 Chr.6: 41061738-41061738 on Build GRCh38
rs374541904 Chr.6: 41061664-41061664 on Build GRCh38
rs771846295 Chr.6: 41061805-41061805 on Build GRCh38

View Full Product Details