Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 50923838-50924107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4E1
Entrez Gene ID
23431
Gene Name
adaptor related protein complex 4 epsilon 1 subunit
Gene Aliases
CPSQ4, SPG51, STUT1
UniGene
Hs.413366
Gene Chromosome Location
Chr.15: 50908569-51005900 on Build GRCh38
Transcripts
NM_001252127.1 NM_007347.4 XM_005254264.3 XM_006720447.3 XM_017022042.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400161 c.347-7T>A p.? Chr.15: 50923924-50923924 on Build GRCh38
470777 c.380G>T p.S127I Chr.15: 50923964-50923964 on Build GRCh38
5383473 c.363C>T p.S121S Chr.15: 50923947-50923947 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779014673 Chr.15: 50924012-50924012 on Build GRCh38
rs144039601 Chr.15: 50923975-50923975 on Build GRCh38
rs749927467 Chr.15: 50923958-50923958 on Build GRCh38
rs745941167 Chr.15: 50924023-50924023 on Build GRCh38
rs766603892 Chr.15: 50923879-50923879 on Build GRCh38
rs777020273 Chr.15: 50924037-50924037 on Build GRCh38
rs764674581 Chr.15: 50923939-50923939 on Build GRCh38
rs775631080 Chr.15: 50924027-50924027 on Build GRCh38
rs761412764 Chr.15: 50923956-50923956 on Build GRCh38
rs560528248 Chr.15: 50924062-50924062 on Build GRCh38
rs1041308162 Chr.15: 50923989-50923989 on Build GRCh38
rs1025797746 Chr.15: 50923897-50923897 on Build GRCh38
rs768858016 Chr.15: 50924034-50924034 on Build GRCh38
rs765810017 Chr.15: 50923923-50923923 on Build GRCh38
rs772284984 Chr.15: 50924024-50924024 on Build GRCh38
rs754675487 Chr.15: 50923983-50923983 on Build GRCh38
rs769741197 Chr.15: 50923999-50923999 on Build GRCh38
rs370227106 Chr.15: 50923957-50923957 on Build GRCh38
rs754250321 Chr.15: 50923945-50923945 on Build GRCh38
rs1053057349 Chr.15: 50923872-50923872 on Build GRCh38
rs779851343 Chr.15: 50923974-50923974 on Build GRCh38
rs762488707 Chr.15: 50924050-50924050 on Build GRCh38
rs762389793 Chr.15: 50923895-50923898 on Build GRCh38
rs995815739 Chr.15: 50924004-50924004 on Build GRCh38
rs758179654 Chr.15: 50923959-50923959 on Build GRCh38
rs774588225 Chr.15: 50923883-50923883 on Build GRCh38
rs192967080 Chr.15: 50924044-50924044 on Build GRCh38
rs781216686 Chr.15: 50923988-50923988 on Build GRCh38
rs747995412 Chr.15: 50923992-50923992 on Build GRCh38
rs370712651 Chr.15: 50924039-50924039 on Build GRCh38
rs772927163 Chr.15: 50924043-50924043 on Build GRCh38
rs373919696 Chr.15: 50924032-50924032 on Build GRCh38
rs571703170 Chr.15: 50924057-50924057 on Build GRCh38
rs760880865 Chr.15: 50923916-50923916 on Build GRCh38
rs775897017 Chr.15: 50923905-50923905 on Build GRCh38
rs1024916938 Chr.15: 50924059-50924059 on Build GRCh38
rs766725563 Chr.15: 50924005-50924009 on Build GRCh38
rs766146810 Chr.15: 50924052-50924052 on Build GRCh38
rs772262339 Chr.15: 50923901-50923901 on Build GRCh38
rs759795254 Chr.15: 50923900-50923900 on Build GRCh38

View Full Product Details