Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 49787280-49787507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAA2
Entrez Gene ID
10449
Gene Name
acetyl-CoA acyltransferase 2
Gene Aliases
DSAEC
UniGene
Hs.200136
Gene Chromosome Location
Chr.18: 49783504-49813881 on Build GRCh38
Transcripts
NM_006111.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6232148 c.907G>A p.A303T Chr.18: 49787338-49787338 on Build GRCh38
1388940 c.896C>T p.A299V Chr.18: 49787349-49787349 on Build GRCh38
6465418 c.925C>T p.L309L Chr.18: 49787320-49787320 on Build GRCh38
7194495 c.898A>G p.I300V Chr.18: 49787347-49787347 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1030536195 Chr.18: 49787466-49787466 on Build GRCh38
rs753857797 Chr.18: 49787373-49787373 on Build GRCh38
rs778963985 Chr.18: 49787408-49787408 on Build GRCh38
rs539036880 Chr.18: 49787309-49787309 on Build GRCh38
rs573418817 Chr.18: 49787376-49787376 on Build GRCh38
rs553645677 Chr.18: 49787368-49787368 on Build GRCh38
rs1016925442 Chr.18: 49787405-49787405 on Build GRCh38
rs186500542 Chr.18: 49787463-49787463 on Build GRCh38
rs969811610 Chr.18: 49787437-49787437 on Build GRCh38
rs995566665 Chr.18: 49787420-49787420 on Build GRCh38
rs767750275 Chr.18: 49787371-49787376 on Build GRCh38
rs755199067 Chr.18: 49787394-49787394 on Build GRCh38
rs536052211 Chr.18: 49787421-49787421 on Build GRCh38
rs558814587 Chr.18: 49787464-49787464 on Build GRCh38
rs1015845306 Chr.18: 49787404-49787404 on Build GRCh38
rs756257897 Chr.18: 49787335-49787335 on Build GRCh38
rs886571161 Chr.18: 49787310-49787310 on Build GRCh38
rs536329631 Chr.18: 49787320-49787320 on Build GRCh38
rs894367911 Chr.18: 49787347-49787347 on Build GRCh38
rs60310353 Chr.18: 49787457-49787457 on Build GRCh38
rs981147956 Chr.18: 49787458-49787458 on Build GRCh38
rs56085706 Chr.18: 49787480-49787480 on Build GRCh38
rs756349396 Chr.18: 49787432-49787434 on Build GRCh38
rs773937153 Chr.18: 49787408-49787408 on Build GRCh38

View Full Product Details