Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46680340-46680567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP1
Entrez Gene ID
392
Gene Name
Rho GTPase activating protein 1
Gene Aliases
CDC42GAP, RHOGAP, RHOGAP1, p50rhoGAP
UniGene
Hs.138860
Gene Chromosome Location
Chr.11: 46677075-46700665 on Build GRCh38
Transcripts
NM_004308.3 XM_011520095.1 XM_017017720.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7307758 c.794A>G p.E265G Chr.11: 46680513-46680513 on Build GRCh38
4033052 c.788T>C p.L263P Chr.11: 46680519-46680519 on Build GRCh38
5838613 c.792_793GG>TT p.R264_E265>S* Chr.11: 46680514-46680515 on Build GRCh38
3752374 c.787C>T p.L263F Chr.11: 46680520-46680520 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752068017 Chr.11: 46680516-46680516 on Build GRCh38
rs755357915 Chr.11: 46680441-46680441 on Build GRCh38
rs765488258 Chr.11: 46680491-46680491 on Build GRCh38
rs748943036 Chr.11: 46680546-46680546 on Build GRCh38
rs777427802 Chr.11: 46680536-46680536 on Build GRCh38
rs368063556 Chr.11: 46680526-46680526 on Build GRCh38
rs767950337 Chr.11: 46680521-46680521 on Build GRCh38
rs899048936 Chr.11: 46680538-46680538 on Build GRCh38
rs756496497 Chr.11: 46680532-46680532 on Build GRCh38
rs981832521 Chr.11: 46680465-46680465 on Build GRCh38
rs774206343 Chr.11: 46680474-46680474 on Build GRCh38
rs867261728 Chr.11: 46680391-46680391 on Build GRCh38
rs954195235 Chr.11: 46680426-46680426 on Build GRCh38
rs777357005 Chr.11: 46680448-46680448 on Build GRCh38
rs763066412 Chr.11: 46680508-46680508 on Build GRCh38
rs139663325 Chr.11: 46680547-46680547 on Build GRCh38
rs748785596 Chr.11: 46680462-46680462 on Build GRCh38
rs917412360 Chr.11: 46680457-46680457 on Build GRCh38
rs144801476 Chr.11: 46680520-46680520 on Build GRCh38
rs989515064 Chr.11: 46680375-46680375 on Build GRCh38
rs532359438 Chr.11: 46680450-46680450 on Build GRCh38
rs745834665 Chr.11: 46680477-46680477 on Build GRCh38
rs371476247 Chr.11: 46680444-46680444 on Build GRCh38
rs376845196 Chr.11: 46680447-46680447 on Build GRCh38
rs766600654 Chr.11: 46680509-46680509 on Build GRCh38
rs544573910 Chr.11: 46680376-46680376 on Build GRCh38
rs771804827 Chr.11: 46680478-46680478 on Build GRCh38
rs373706197 Chr.11: 46680464-46680464 on Build GRCh38
rs112395124 Chr.11: 46680443-46680443 on Build GRCh38
rs773361751 Chr.11: 46680505-46680505 on Build GRCh38
rs530673495 Chr.11: 46680419-46680419 on Build GRCh38
rs564837812 Chr.11: 46680415-46680415 on Build GRCh38
rs760424122 Chr.11: 46680487-46680487 on Build GRCh38

View Full Product Details