Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46644189-46644462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
UniGene
Hs.127403
Gene Chromosome Location
Chr.11: 46617276-46676019 on Build GRCh38
Transcripts
NM_001142673.2 NM_001205119.1 NM_001205120.1 NM_001205121.1 NM_001205122.1 NM_014741.4 XM_005253262.2 XM_005253263.2 XM_005253265.2 XM_005253266.2 XM_005253268.2 XM_006718394.2 XM_006718395.2 XM_006718396.2 XM_011520493.1 XM_011520496.2 XM_011520499.2 XM_011520500.1 XM_011520501.1 XM_011520502.1 XM_011520503.1 XM_017018599.1 XM_017018600.1 XM_017018601.1 XM_017018602.1 XM_017018603.1 XM_017018604.1 XM_017018605.1 XM_017018606.1 XM_017018607.1 XM_017018608.1 XM_017018609.1 XM_017018610.1 XM_017018611.1 XM_017018612.1 XM_017018613.1 XM_017018614.1 XM_017018615.1 XM_017018616.1 XM_017018617.1 XM_017018618.1 XM_017018619.1 XM_017018620.1 XM_017018621.1 XM_017018622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4033041 c.56T>G p.F19C Chr.11: 46644347-46644347 on Build GRCh38
4503443 c.63C>T p.A21A Chr.11: 46644354-46644354 on Build GRCh38
5838610 c.69+1G>A p.? Chr.11: 46644361-46644361 on Build GRCh38
6882716 c.40G>A p.D14N Chr.11: 46644331-46644331 on Build GRCh38
927244 c.67A>G p.K23E Chr.11: 46644358-46644358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376775323 Chr.11: 46644424-46644424 on Build GRCh38
rs988363113 Chr.11: 46644283-46644283 on Build GRCh38
rs755647056 Chr.11: 46644315-46644315 on Build GRCh38
rs774677172 Chr.11: 46644269-46644270 on Build GRCh38
rs573939072 Chr.11: 46644427-46644427 on Build GRCh38
rs748243388 Chr.11: 46644277-46644277 on Build GRCh38
rs763415225 Chr.11: 46644281-46644281 on Build GRCh38
rs148400859 Chr.11: 46644318-46644318 on Build GRCh38
rs747191082 Chr.11: 46644246-46644246 on Build GRCh38
rs750231463 Chr.11: 46644366-46644366 on Build GRCh38
rs778611963 Chr.11: 46644357-46644357 on Build GRCh38
rs759200846 Chr.11: 46644300-46644300 on Build GRCh38
rs969395216 Chr.11: 46644229-46644229 on Build GRCh38
rs763417592 Chr.11: 46644372-46644372 on Build GRCh38
rs887737488 Chr.11: 46644435-46644435 on Build GRCh38
rs753793005 Chr.11: 46644324-46644324 on Build GRCh38
rs1040120075 Chr.11: 46644309-46644309 on Build GRCh38
rs890908933 Chr.11: 46644365-46644365 on Build GRCh38
rs767714532 Chr.11: 46644397-46644405 on Build GRCh38
rs762258154 Chr.11: 46644300-46644301 on Build GRCh38
rs748354792 Chr.11: 46644392-46644392 on Build GRCh38
rs781469080 Chr.11: 46644381-46644381 on Build GRCh38
rs774892074 Chr.11: 46644291-46644291 on Build GRCh38
rs752352332 Chr.11: 46644308-46644308 on Build GRCh38
rs772712601 Chr.11: 46644408-46644408 on Build GRCh38
rs759095489 Chr.11: 46644266-46644267 on Build GRCh38
rs73451860 Chr.11: 46644249-46644249 on Build GRCh38
rs372562738 Chr.11: 46644407-46644407 on Build GRCh38
rs770702341 Chr.11: 46644266-46644266 on Build GRCh38
rs746591479 Chr.11: 46644255-46644256 on Build GRCh38
rs773734147 Chr.11: 46644280-46644280 on Build GRCh38
rs750958461 Chr.11: 46644265-46644266 on Build GRCh38
rs144644606 Chr.11: 46644304-46644304 on Build GRCh38
rs769829577 Chr.11: 46644279-46644279 on Build GRCh38
rs750851624 Chr.11: 46644355-46644355 on Build GRCh38
rs766804955 Chr.11: 46644285-46644285 on Build GRCh38
rs768868930 Chr.11: 46644247-46644247 on Build GRCh38
rs757216241 Chr.11: 46644332-46644332 on Build GRCh38

View Full Product Details