Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154588476-154588749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAR
Entrez Gene ID
103
Gene Name
adenosine deaminase, RNA specific
Gene Aliases
ADAR1, AGS6, DRADA, DSH, DSRAD, G1P1, IFI-4, IFI4, K88DSRBP, P136
UniGene
Hs.12341
Gene Chromosome Location
Chr.1: 154582057-154628016 on Build GRCh38
Transcripts
NM_001025107.2 NM_001111.4 NM_001193495.1 NM_015840.3 NM_015841.3 XM_006711109.2 XM_006711111.3 XM_006711112.2 XM_006711113.2 XM_011509060.1 XM_011509061.2 XM_011509062.1 XM_017000037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4022837 c.2987A>G p.K996R Chr.1: 154588578-154588578 on Build GRCh38
4791360 c.2945T>C p.I982T Chr.1: 154588620-154588620 on Build GRCh38
5341565 c.2832G>A p.K944K Chr.1: 154588604-154588604 on Build GRCh38
1334459 c.2804C>T p.A935V Chr.1: 154588632-154588632 on Build GRCh38
6121521 c.2879A>G p.Y960C Chr.1: 154588557-154588557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769300917 Chr.1: 154588563-154588563 on Build GRCh38
rs370087378 Chr.1: 154588538-154588538 on Build GRCh38
rs373564780 Chr.1: 154588606-154588606 on Build GRCh38
rs764167064 Chr.1: 154588669-154588669 on Build GRCh38
rs928347941 Chr.1: 154588539-154588539 on Build GRCh38
rs763453697 Chr.1: 154588604-154588604 on Build GRCh38
rs779762646 Chr.1: 154588720-154588720 on Build GRCh38
rs746643040 Chr.1: 154588658-154588658 on Build GRCh38
rs886082285 Chr.1: 154588641-154588641 on Build GRCh38
rs144280682 Chr.1: 154588504-154588504 on Build GRCh38
rs747691527 Chr.1: 154588552-154588552 on Build GRCh38
rs368855871 Chr.1: 154588716-154588716 on Build GRCh38
rs112563989 Chr.1: 154588550-154588550 on Build GRCh38
rs1046161321 Chr.1: 154588637-154588637 on Build GRCh38
rs778736829 Chr.1: 154588524-154588524 on Build GRCh38
rs758131055 Chr.1: 154588533-154588533 on Build GRCh38
rs773609252 Chr.1: 154588578-154588578 on Build GRCh38
rs967132792 Chr.1: 154588535-154588535 on Build GRCh38
rs752917999 Chr.1: 154588652-154588652 on Build GRCh38
rs759815056 Chr.1: 154588513-154588513 on Build GRCh38
rs121912422 Chr.1: 154588582-154588582 on Build GRCh38
rs777215669 Chr.1: 154588611-154588611 on Build GRCh38
rs780722178 Chr.1: 154588548-154588548 on Build GRCh38
rs375643914 Chr.1: 154588631-154588631 on Build GRCh38
rs768277709 Chr.1: 154588540-154588540 on Build GRCh38
rs750316347 Chr.1: 154588527-154588527 on Build GRCh38
rs762694073 Chr.1: 154588532-154588533 on Build GRCh38
rs752789655 Chr.1: 154588517-154588517 on Build GRCh38
rs28936680 Chr.1: 154588668-154588668 on Build GRCh38
rs536855590 Chr.1: 154588537-154588537 on Build GRCh38
rs1054969472 Chr.1: 154588612-154588612 on Build GRCh38
rs767879893 Chr.1: 154588632-154588632 on Build GRCh38
rs375555599 Chr.1: 154588529-154588529 on Build GRCh38
rs750394212 Chr.1: 154588683-154588683 on Build GRCh38
rs756221691 Chr.1: 154588521-154588521 on Build GRCh38
rs368948193 Chr.1: 154588667-154588667 on Build GRCh38
rs770539977 Chr.1: 154588719-154588719 on Build GRCh38
rs767752404 Chr.1: 154588514-154588514 on Build GRCh38
rs774830798 Chr.1: 154588621-154588621 on Build GRCh38

View Full Product Details