Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1041587-1041852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA7
Entrez Gene ID
10347
Gene Name
ATP binding cassette subfamily A member 7
Gene Aliases
ABCA-SSN, ABCX, AD9
UniGene
Hs.134514
Gene Chromosome Location
Chr.19: 1040103-1065572 on Build GRCh38
Transcripts
NM_019112.3 XM_006722616.1 XM_006722617.2 XM_011527628.2 XM_011527629.1 XM_011527630.1 XM_011527631.1 XM_011527633.2 XM_011527634.1 XM_011527635.1 XM_017026142.1 XM_017026143.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2151645 c.174C>A p.N58K Chr.19: 1041844-1041844 on Build GRCh38
6589822 c.160+6delC p.? Chr.19: 1041609-1041609 on Build GRCh38
3528151 c.148G>C p.E50Q Chr.19: 1041591-1041591 on Build GRCh38
6589834 c.160+5_160+6insC p.? Chr.19: 1041608-1041609 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761645910 Chr.19: 1041653-1041653 on Build GRCh38
rs757699680 Chr.19: 1041819-1041819 on Build GRCh38
rs1048417153 Chr.19: 1041802-1041802 on Build GRCh38
rs767415887 Chr.19: 1041785-1041785 on Build GRCh38
rs764668443 Chr.19: 1041814-1041814 on Build GRCh38
rs568239562 Chr.19: 1041610-1041610 on Build GRCh38
rs756495449 Chr.19: 1041826-1041826 on Build GRCh38
rs566014616 Chr.19: 1041776-1041776 on Build GRCh38
rs547618420 Chr.19: 1041763-1041763 on Build GRCh38
rs766043989 Chr.19: 1041808-1041808 on Build GRCh38
rs778841642 Chr.19: 1041609-1041609 on Build GRCh38
rs977899456 Chr.19: 1041707-1041707 on Build GRCh38
rs199601811 Chr.19: 1041827-1041827 on Build GRCh38
rs768813299 Chr.19: 1041647-1041647 on Build GRCh38
rs879178591 Chr.19: 1041663-1041663 on Build GRCh38
rs746192296 Chr.19: 1041823-1041823 on Build GRCh38
rs772854996 Chr.19: 1041787-1041787 on Build GRCh38
rs762408445 Chr.19: 1041622-1041622 on Build GRCh38
rs770109695 Chr.19: 1041616-1041616 on Build GRCh38
rs746167040 Chr.19: 1041614-1041614 on Build GRCh38
rs948732082 Chr.19: 1041780-1041780 on Build GRCh38
rs567043891 Chr.19: 1041716-1041716 on Build GRCh38
rs370037870 Chr.19: 1041809-1041809 on Build GRCh38
rs751125068 Chr.19: 1041764-1041764 on Build GRCh38
rs939257344 Chr.19: 1041758-1041758 on Build GRCh38
rs774432275 Chr.19: 1041649-1041649 on Build GRCh38
rs768917670 Chr.19: 1041829-1041829 on Build GRCh38
rs557727395 Chr.19: 1041812-1041812 on Build GRCh38
rs981273963 Chr.19: 1041722-1041723 on Build GRCh38
rs760429218 Chr.19: 1041795-1041795 on Build GRCh38
rs775757250 Chr.19: 1041621-1041621 on Build GRCh38
rs749397103 Chr.19: 1041644-1041644 on Build GRCh38
rs918490906 Chr.19: 1041767-1041767 on Build GRCh38
rs367574717 Chr.19: 1041613-1041613 on Build GRCh38
rs927813382 Chr.19: 1041724-1041724 on Build GRCh38
rs781650490 Chr.19: 1041821-1041821 on Build GRCh38
rs754757371 Chr.19: 1041608-1041609 on Build GRCh38

View Full Product Details