Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 38595790-38596061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD18A
Entrez Gene ID
253650
Gene Name
ankyrin repeat domain 18A
Gene Aliases
-
UniGene
Hs.644630
Gene Chromosome Location
Chr.9: 38566260-38620587 on Build GRCh38
Transcripts
NM_147195.2 XM_006716749.3 XM_011517836.2 XM_011517837.2 XM_011517839.2 XM_017014576.1 XM_017014577.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1177806 c.1420G>A p.E474K Chr.9: 38595920-38595920 on Build GRCh38
5893340 c.1474G>A p.E492K Chr.9: 38595866-38595866 on Build GRCh38
5506112 c.1472G>A p.R491H Chr.9: 38595868-38595868 on Build GRCh38
6260366 c.1323C>A p.D441E Chr.9: 38596017-38596017 on Build GRCh38
5416285 c.1303G>T p.E435* Chr.9: 38596037-38596037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2996347 Chr.9: 38595889-38595889 on Build GRCh38
rs544352530 Chr.9: 38595975-38595975 on Build GRCh38
rs991207820 Chr.9: 38595970-38595970 on Build GRCh38
rs752527220 Chr.9: 38595872-38595872 on Build GRCh38
rs746349646 Chr.9: 38596016-38596016 on Build GRCh38
rs759573339 Chr.9: 38595855-38595855 on Build GRCh38
rs920443747 Chr.9: 38595924-38595924 on Build GRCh38
rs573170797 Chr.9: 38595813-38595813 on Build GRCh38
rs777992111 Chr.9: 38595933-38595933 on Build GRCh38
rs779117794 Chr.9: 38596001-38596001 on Build GRCh38
rs757289252 Chr.9: 38595991-38595991 on Build GRCh38
rs187514653 Chr.9: 38595918-38595918 on Build GRCh38
rs955363092 Chr.9: 38595818-38595818 on Build GRCh38
rs200079271 Chr.9: 38595902-38595902 on Build GRCh38
rs958538006 Chr.9: 38595935-38595935 on Build GRCh38
rs1029716726 Chr.9: 38595810-38595810 on Build GRCh38
rs770173970 Chr.9: 38595984-38595984 on Build GRCh38
rs540395361 Chr.9: 38595823-38595823 on Build GRCh38
rs111615247 Chr.9: 38595880-38595880 on Build GRCh38
rs753857974 Chr.9: 38595964-38595964 on Build GRCh38
rs561815251 Chr.9: 38595868-38595868 on Build GRCh38

View Full Product Details