Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30912049-30912306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP1
Entrez Gene ID
358
Gene Name
aquaporin 1 (Colton blood group)
Gene Aliases
AQP-CHIP, CHIP28, CO
UniGene
Hs.76152
Gene Chromosome Location
Chr.7: 30911800-30925516 on Build GRCh38
Transcripts
NM_198098.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6020489 c.338C>G p.S113* Chr.7: 30912247-30912247 on Build GRCh38
746197 c.377G>A p.R126H Chr.7: 30912286-30912286 on Build GRCh38
4435957 c.186G>A p.T62T Chr.7: 30912095-30912095 on Build GRCh38
318738 c.384+1G>A p.? Chr.7: 30912294-30912294 on Build GRCh38
6177671 c.307G>T p.V103L Chr.7: 30912216-30912216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898407370 Chr.7: 30912100-30912100 on Build GRCh38
rs183830991 Chr.7: 30912187-30912187 on Build GRCh38
rs771986400 Chr.7: 30912175-30912175 on Build GRCh38
rs753051699 Chr.7: 30912090-30912090 on Build GRCh38
rs372063183 Chr.7: 30912192-30912192 on Build GRCh38
rs759042117 Chr.7: 30912196-30912196 on Build GRCh38
rs144761239 Chr.7: 30912215-30912215 on Build GRCh38
rs762474222 Chr.7: 30912216-30912216 on Build GRCh38
rs968619407 Chr.7: 30912112-30912112 on Build GRCh38
rs995925781 Chr.7: 30912278-30912278 on Build GRCh38
rs35804488 Chr.7: 30912228-30912228 on Build GRCh38
rs140979736 Chr.7: 30912126-30912126 on Build GRCh38
rs768291072 Chr.7: 30912206-30912206 on Build GRCh38
rs769066207 Chr.7: 30912273-30912273 on Build GRCh38
rs556371635 Chr.7: 30912286-30912286 on Build GRCh38
rs749425186 Chr.7: 30912128-30912128 on Build GRCh38
rs768530306 Chr.7: 30912139-30912139 on Build GRCh38
rs940136659 Chr.7: 30912231-30912231 on Build GRCh38
rs757778816 Chr.7: 30912115-30912115 on Build GRCh38
rs746813756 Chr.7: 30912194-30912194 on Build GRCh38
rs758373056 Chr.7: 30912240-30912240 on Build GRCh38
rs199545971 Chr.7: 30912094-30912094 on Build GRCh38
rs368313004 Chr.7: 30912095-30912095 on Build GRCh38
rs761024461 Chr.7: 30912287-30912287 on Build GRCh38
rs368861831 Chr.7: 30912230-30912230 on Build GRCh38
rs550724674 Chr.7: 30912077-30912077 on Build GRCh38
rs772776753 Chr.7: 30912182-30912182 on Build GRCh38
rs778827593 Chr.7: 30912237-30912237 on Build GRCh38
rs200596948 Chr.7: 30912068-30912068 on Build GRCh38
rs777709519 Chr.7: 30912241-30912241 on Build GRCh38
rs143467679 Chr.7: 30912252-30912252 on Build GRCh38
rs918080655 Chr.7: 30912226-30912226 on Build GRCh38
rs147400857 Chr.7: 30912229-30912229 on Build GRCh38
rs957051800 Chr.7: 30912177-30912177 on Build GRCh38
rs748092933 Chr.7: 30912144-30912144 on Build GRCh38
rs925279000 Chr.7: 30912161-30912161 on Build GRCh38
rs762241529 Chr.7: 30912275-30912275 on Build GRCh38
rs558089401 Chr.7: 30912103-30912103 on Build GRCh38
rs548913885 Chr.7: 30912123-30912123 on Build GRCh38
rs375901301 Chr.7: 30912239-30912239 on Build GRCh38
rs769323781 Chr.7: 30912257-30912257 on Build GRCh38
rs971080900 Chr.7: 30912132-30912132 on Build GRCh38
rs752109788 Chr.7: 30912101-30912101 on Build GRCh38
rs746907533 Chr.7: 30912242-30912242 on Build GRCh38
rs371290302 Chr.7: 30912236-30912236 on Build GRCh38
rs770477413 Chr.7: 30912250-30912250 on Build GRCh38
rs112240167 Chr.7: 30912140-30912140 on Build GRCh38
rs767949083 Chr.7: 30912277-30912277 on Build GRCh38
rs745590240 Chr.7: 30912256-30912256 on Build GRCh38
rs145002350 Chr.7: 30912125-30912125 on Build GRCh38
rs375712827 Chr.7: 30912227-30912227 on Build GRCh38
rs11537656 Chr.7: 30912089-30912089 on Build GRCh38
rs954729721 Chr.7: 30912069-30912069 on Build GRCh38
rs760146829 Chr.7: 30912186-30912186 on Build GRCh38
rs774892365 Chr.7: 30912274-30912274 on Build GRCh38

View Full Product Details