Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151756938-151757173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADACL2
Entrez Gene ID
344752
Gene Name
arylacetamide deacetylase like 2
Gene Aliases
-
UniGene
Hs.144710
Gene Chromosome Location
Chr.3: 151733916-151758016 on Build GRCh38
Transcripts
NM_207365.3 XM_011512753.1 NR_110202.1 NR_110203.1

Gene Symbol
AADACL2-AS1
Entrez Gene ID
101928142
Gene Name
AADACL2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.680207
Gene Chromosome Location
Chr.3: 151751179-151928175 on Build GRCh38
Transcripts
NM_207365.3 XM_011512753.1 NR_110202.1 NR_110203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6096792 c.549T>C p.D183D Chr.3: 151757003-151757003 on Build GRCh38
6163764 c.659C>A p.T220N Chr.3: 151757113-151757113 on Build GRCh38
1752973 c.668T>C p.V223A Chr.3: 151757122-151757122 on Build GRCh38
131313 c.682G>T p.V228L Chr.3: 151757136-151757136 on Build GRCh38
5474566 c.631C>T p.R211* Chr.3: 151757085-151757085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757373885 Chr.3: 151757099-151757099 on Build GRCh38
rs896654043 Chr.3: 151757111-151757111 on Build GRCh38
rs759026153 Chr.3: 151757106-151757106 on Build GRCh38
rs763731114 Chr.3: 151757086-151757086 on Build GRCh38
rs933697412 Chr.3: 151756985-151756985 on Build GRCh38
rs765538270 Chr.3: 151757139-151757139 on Build GRCh38
rs575496882 Chr.3: 151757075-151757075 on Build GRCh38
rs17855338 Chr.3: 151757033-151757033 on Build GRCh38
rs769393189 Chr.3: 151757011-151757011 on Build GRCh38
rs773612462 Chr.3: 151757069-151757069 on Build GRCh38
rs772910203 Chr.3: 151757031-151757031 on Build GRCh38
rs371561457 Chr.3: 151757062-151757062 on Build GRCh38
rs747396499 Chr.3: 151757114-151757114 on Build GRCh38
rs905472160 Chr.3: 151757094-151757094 on Build GRCh38
rs150410369 Chr.3: 151757151-151757151 on Build GRCh38
rs1056622497 Chr.3: 151757104-151757104 on Build GRCh38
rs375567161 Chr.3: 151757070-151757070 on Build GRCh38
rs754104886 Chr.3: 151757096-151757096 on Build GRCh38
rs987917550 Chr.3: 151757038-151757038 on Build GRCh38
rs77715518 Chr.3: 151757085-151757085 on Build GRCh38
rs757315486 Chr.3: 151757138-151757138 on Build GRCh38
rs779169325 Chr.3: 151757101-151757101 on Build GRCh38
rs369501242 Chr.3: 151757136-151757136 on Build GRCh38
rs975189839 Chr.3: 151756964-151756964 on Build GRCh38
rs747379679 Chr.3: 151756996-151756996 on Build GRCh38
rs760473210 Chr.3: 151757135-151757135 on Build GRCh38
rs748987722 Chr.3: 151757116-151757116 on Build GRCh38
rs767498608 Chr.3: 151757126-151757126 on Build GRCh38
rs375687513 Chr.3: 151757103-151757103 on Build GRCh38
rs772671335 Chr.3: 151756981-151756981 on Build GRCh38
rs180742060 Chr.3: 151756980-151756980 on Build GRCh38
rs768328573 Chr.3: 151757062-151757063 on Build GRCh38
rs762443164 Chr.3: 151757039-151757039 on Build GRCh38
rs993954231 Chr.3: 151757113-151757113 on Build GRCh38
rs201772248 Chr.3: 151756965-151756965 on Build GRCh38
rs750591739 Chr.3: 151757143-151757143 on Build GRCh38
rs868684290 Chr.3: 151757147-151757147 on Build GRCh38
rs776800077 Chr.3: 151757115-151757115 on Build GRCh38
rs765922865 Chr.3: 151757045-151757045 on Build GRCh38
rs770531172 Chr.3: 151757118-151757118 on Build GRCh38
rs753536191 Chr.3: 151757137-151757137 on Build GRCh38
rs773890145 Chr.3: 151757119-151757119 on Build GRCh38
rs186045529 Chr.3: 151756982-151756982 on Build GRCh38
rs780696160 Chr.3: 151757112-151757112 on Build GRCh38
rs759022730 Chr.3: 151757121-151757121 on Build GRCh38
rs368531994 Chr.3: 151757072-151757072 on Build GRCh38
rs773798017 Chr.3: 151757056-151757056 on Build GRCh38
rs913551817 Chr.3: 151757050-151757050 on Build GRCh38
rs974273719 Chr.3: 151757059-151757059 on Build GRCh38
rs140496674 Chr.3: 151757081-151757081 on Build GRCh38
rs779079275 Chr.3: 151756966-151756966 on Build GRCh38
rs201138455 Chr.3: 151757129-151757129 on Build GRCh38

View Full Product Details