Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81899853-81900112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC11
Entrez Gene ID
51529
Gene Name
anaphase promoting complex subunit 11
Gene Aliases
APC11, Apc11p, HSPC214
UniGene
Hs.534456
Gene Chromosome Location
Chr.17: 81890790-81900533 on Build GRCh38
Transcripts
NM_001002244.2 NM_001002245.2 NM_001002246.2 NM_001002247.2 NM_001002248.2 NM_001002249.2 NM_001289414.1 NM_001289415.1 NM_001289416.1 NM_001289417.1 NM_001289418.1 NM_001289419.1 NM_001289420.1 NM_016476.11

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6399261 c.414A>T p.Q138H Chr.17: 81899923-81899923 on Build GRCh38
6082526 c.428G>T p.R143L Chr.17: 81899937-81899937 on Build GRCh38
4608696 c.493G>T p.A165S Chr.17: 81900002-81900002 on Build GRCh38
6599523 c.497G>A p.R166H Chr.17: 81900006-81900006 on Build GRCh38
7292013 c.424C>T p.R142* Chr.17: 81899933-81899933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767273965 Chr.17: 81899945-81899945 on Build GRCh38
rs747246909 Chr.17: 81900062-81900062 on Build GRCh38
rs924392175 Chr.17: 81900006-81900006 on Build GRCh38
rs756571746 Chr.17: 81900023-81900023 on Build GRCh38
rs760002984 Chr.17: 81899933-81899933 on Build GRCh38
rs754818707 Chr.17: 81900005-81900005 on Build GRCh38
rs549791007 Chr.17: 81899891-81899891 on Build GRCh38
rs886460643 Chr.17: 81900090-81900090 on Build GRCh38
rs149063288 Chr.17: 81899963-81899963 on Build GRCh38
rs752711149 Chr.17: 81900039-81900039 on Build GRCh38
rs771296893 Chr.17: 81900070-81900070 on Build GRCh38
rs770357311 Chr.17: 81900085-81900085 on Build GRCh38
rs933574096 Chr.17: 81899879-81899879 on Build GRCh38
rs1026342272 Chr.17: 81900082-81900082 on Build GRCh38
rs372436095 Chr.17: 81899895-81899895 on Build GRCh38
rs777973315 Chr.17: 81899916-81899916 on Build GRCh38
rs565397041 Chr.17: 81899942-81899942 on Build GRCh38
rs535288823 Chr.17: 81899931-81899931 on Build GRCh38
rs767292388 Chr.17: 81899886-81899886 on Build GRCh38
rs776205373 Chr.17: 81900091-81900091 on Build GRCh38
rs368182201 Chr.17: 81899876-81899876 on Build GRCh38
rs375206392 Chr.17: 81899915-81899915 on Build GRCh38
rs143094452 Chr.17: 81899966-81899966 on Build GRCh38
rs749235863 Chr.17: 81899896-81899896 on Build GRCh38
rs778838908 Chr.17: 81900026-81900026 on Build GRCh38
rs746266905 Chr.17: 81900083-81900083 on Build GRCh38
rs1002782689 Chr.17: 81900080-81900080 on Build GRCh38
rs761412036 Chr.17: 81899944-81899944 on Build GRCh38
rs11550402 Chr.17: 81900042-81900042 on Build GRCh38
rs557305511 Chr.17: 81900087-81900087 on Build GRCh38
rs781372181 Chr.17: 81900071-81900071 on Build GRCh38
rs34978095 Chr.17: 81899952-81899953 on Build GRCh38
rs755057760 Chr.17: 81899901-81899901 on Build GRCh38
rs865840307 Chr.17: 81900077-81900077 on Build GRCh38
rs777961227 Chr.17: 81900055-81900055 on Build GRCh38
rs539064554 Chr.17: 81899948-81899948 on Build GRCh38
rs531665657 Chr.17: 81899883-81899883 on Build GRCh38
rs778928428 Chr.17: 81899910-81899910 on Build GRCh38
rs753694115 Chr.17: 81899982-81899982 on Build GRCh38
rs137864015 Chr.17: 81899936-81899936 on Build GRCh38
rs747366718 Chr.17: 81899920-81899920 on Build GRCh38
rs748254134 Chr.17: 81899914-81899914 on Build GRCh38
rs758543327 Chr.17: 81900047-81900047 on Build GRCh38
rs11550400 Chr.17: 81900024-81900024 on Build GRCh38
rs771255547 Chr.17: 81899930-81899930 on Build GRCh38

View Full Product Details