Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50192428-50192677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP4
Entrez Gene ID
8852
Gene Name
A-kinase anchoring protein 4
Gene Aliases
AKAP 82, AKAP-4, AKAP82, CT99, FSC1, HI, PRKA4, hAKAP82, p82
UniGene
Hs.97633
Gene Chromosome Location
Chr.X: 50190768-50201013 on Build GRCh38
Transcripts
NM_003886.2 NM_139289.1

SNP Detail

SNP ID Chromosome Location
rs145663575 Chr.X: 50192523-50192523 on Build GRCh38
rs138199562 Chr.X: 50192557-50192557 on Build GRCh38
rs782653772 Chr.X: 50192522-50192522 on Build GRCh38
rs12011833 Chr.X: 50192565-50192565 on Build GRCh38
rs782216204 Chr.X: 50192530-50192530 on Build GRCh38
rs61751433 Chr.X: 50192509-50192509 on Build GRCh38
rs782053667 Chr.X: 50192568-50192568 on Build GRCh38
rs782721380 Chr.X: 50192489-50192489 on Build GRCh38
rs781868791 Chr.X: 50192500-50192500 on Build GRCh38
rs781886679 Chr.X: 50192514-50192514 on Build GRCh38
rs374992303 Chr.X: 50192614-50192614 on Build GRCh38
rs782630376 Chr.X: 50192552-50192552 on Build GRCh38
rs1047259655 Chr.X: 50192582-50192582 on Build GRCh38
rs781791898 Chr.X: 50192470-50192470 on Build GRCh38
rs782773415 Chr.X: 50192648-50192648 on Build GRCh38
rs370785257 Chr.X: 50192463-50192463 on Build GRCh38
rs782127705 Chr.X: 50192638-50192638 on Build GRCh38
rs530483931 Chr.X: 50192502-50192502 on Build GRCh38
rs782291865 Chr.X: 50192488-50192488 on Build GRCh38
rs782296919 Chr.X: 50192555-50192555 on Build GRCh38

View Full Product Details