Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7700660-7700933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WRAP53
Entrez Gene ID
55135
Gene Name
WD repeat containing antisense to TP53
Gene Aliases
DKCB3, TCAB1, WDR79
UniGene
Hs.408312
Gene Chromosome Location
Chr.17: 7686071-7703502 on Build GRCh38
Transcripts
NM_001143990.1 NM_001143991.1 NM_001143992.1 NM_018081.2 XM_011523952.2 XM_017024812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2745210 c.652C>T p.L218F Chr.17: 7700750-7700750 on Build GRCh38
4838093 c.696T>A p.Y232* Chr.17: 7700794-7700794 on Build GRCh38
7397937 c.646C>T p.P216S Chr.17: 7700744-7700744 on Build GRCh38
3522719 c.655C>T p.R219* Chr.17: 7700753-7700753 on Build GRCh38
6262034 c.649G>T p.V217F Chr.17: 7700747-7700747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765264100 Chr.17: 7700768-7700768 on Build GRCh38
rs866951873 Chr.17: 7700749-7700749 on Build GRCh38
rs760316057 Chr.17: 7700707-7700707 on Build GRCh38
rs773908331 Chr.17: 7700753-7700753 on Build GRCh38
rs770005848 Chr.17: 7700737-7700737 on Build GRCh38
rs767803897 Chr.17: 7700778-7700778 on Build GRCh38
rs753668696 Chr.17: 7700766-7700766 on Build GRCh38
rs753790148 Chr.17: 7700721-7700721 on Build GRCh38
rs527734640 Chr.17: 7700809-7700809 on Build GRCh38
rs776637813 Chr.17: 7700850-7700850 on Build GRCh38
rs755312840 Chr.17: 7700780-7700780 on Build GRCh38
rs370637813 Chr.17: 7700686-7700686 on Build GRCh38
rs751679850 Chr.17: 7700726-7700726 on Build GRCh38
rs115388376 Chr.17: 7700865-7700865 on Build GRCh38
rs770755456 Chr.17: 7700907-7700907 on Build GRCh38
rs566624166 Chr.17: 7700780-7700780 on Build GRCh38
rs1024600581 Chr.17: 7700697-7700697 on Build GRCh38
rs138634236 Chr.17: 7700856-7700856 on Build GRCh38
rs570343418 Chr.17: 7700827-7700827 on Build GRCh38
rs751696137 Chr.17: 7700770-7700770 on Build GRCh38
rs749638410 Chr.17: 7700805-7700805 on Build GRCh38
rs757234887 Chr.17: 7700723-7700723 on Build GRCh38
rs754956678 Chr.17: 7700775-7700775 on Build GRCh38
rs1049568496 Chr.17: 7700898-7700898 on Build GRCh38
rs773235897 Chr.17: 7700855-7700855 on Build GRCh38
rs763839659 Chr.17: 7700765-7700765 on Build GRCh38
rs937756256 Chr.17: 7700735-7700735 on Build GRCh38
rs756347392 Chr.17: 7700860-7700860 on Build GRCh38
rs746693191 Chr.17: 7700823-7700823 on Build GRCh38
rs556563734 Chr.17: 7700858-7700858 on Build GRCh38
rs764102509 Chr.17: 7700719-7700719 on Build GRCh38
rs771403969 Chr.17: 7700750-7700750 on Build GRCh38
rs960056891 Chr.17: 7700864-7700864 on Build GRCh38
rs771431030 Chr.17: 7700821-7700821 on Build GRCh38
rs774944601 Chr.17: 7700757-7700757 on Build GRCh38
rs759039161 Chr.17: 7700754-7700754 on Build GRCh38
rs909665404 Chr.17: 7700683-7700683 on Build GRCh38
rs368989809 Chr.17: 7700728-7700728 on Build GRCh38
rs775038177 Chr.17: 7700822-7700822 on Build GRCh38
rs761634556 Chr.17: 7700851-7700851 on Build GRCh38
rs778159976 Chr.17: 7700798-7700798 on Build GRCh38
rs371865260 Chr.17: 7700756-7700756 on Build GRCh38
rs755295065 Chr.17: 7700727-7700727 on Build GRCh38
rs779323513 Chr.17: 7700814-7700814 on Build GRCh38
rs376763835 Chr.17: 7700685-7700685 on Build GRCh38
rs539059271 Chr.17: 7700882-7700882 on Build GRCh38
rs933781000 Chr.17: 7700877-7700877 on Build GRCh38
rs942326138 Chr.17: 7700684-7700684 on Build GRCh38
rs750995622 Chr.17: 7700680-7700680 on Build GRCh38
rs754204126 Chr.17: 7700869-7700869 on Build GRCh38
rs199901527 Chr.17: 7700818-7700818 on Build GRCh38
rs748560120 Chr.17: 7700729-7700729 on Build GRCh38
rs756328444 Chr.17: 7700782-7700782 on Build GRCh38
rs773685166 Chr.17: 7700857-7700857 on Build GRCh38
rs760537751 Chr.17: 7700761-7700761 on Build GRCh38
rs749833877 Chr.17: 7700744-7700744 on Build GRCh38
rs147151745 Chr.17: 7700724-7700724 on Build GRCh38
rs62062588 Chr.17: 7700694-7700694 on Build GRCh38
rs971965334 Chr.17: 7700701-7700701 on Build GRCh38
rs111998875 Chr.17: 7700854-7700854 on Build GRCh38

View Full Product Details