Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 111808880-111809113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC1
Entrez Gene ID
64682
Gene Name
anaphase promoting complex subunit 1
Gene Aliases
APC1, MCPR, TSG24
UniGene
Hs.436527
Gene Chromosome Location
Chr.2: 111766147-111884196 on Build GRCh38
Transcripts
NM_022662.3 XM_011511633.2 XM_011511634.2 XM_017004710.1 XM_017004711.1 XM_017004712.1 XM_017004713.1 XM_017004714.1 XM_017004715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
148927 c.3832+7G>A p.? Chr.2: 111808940-111808940 on Build GRCh38
4084187 c.3677G>A p.R1226Q Chr.2: 111809102-111809102 on Build GRCh38
3565262 c.3680T>A p.L1227H Chr.2: 111809099-111809099 on Build GRCh38
4439878 c.3716C>T p.T1239M Chr.2: 111809063-111809063 on Build GRCh38
5994059 c.3718T>C p.S1240P Chr.2: 111809061-111809061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769388456 Chr.2: 111809067-111809067 on Build GRCh38
rs988269344 Chr.2: 111808908-111808908 on Build GRCh38
rs772884902 Chr.2: 111809069-111809069 on Build GRCh38
rs140589913 Chr.2: 111809074-111809074 on Build GRCh38
rs761362878 Chr.2: 111809064-111809064 on Build GRCh38
rs915012577 Chr.2: 111808909-111808909 on Build GRCh38
rs776088773 Chr.2: 111809063-111809063 on Build GRCh38
rs548389443 Chr.2: 111809002-111809002 on Build GRCh38
rs2686027 Chr.2: 111809087-111809087 on Build GRCh38
rs147822390 Chr.2: 111809079-111809079 on Build GRCh38
rs772625284 Chr.2: 111809042-111809042 on Build GRCh38
rs968816115 Chr.2: 111808917-111808917 on Build GRCh38
rs201128688 Chr.2: 111808940-111808940 on Build GRCh38
rs937632916 Chr.2: 111809055-111809055 on Build GRCh38
rs1036816528 Chr.2: 111809070-111809070 on Build GRCh38
rs927539848 Chr.2: 111809012-111809012 on Build GRCh38
rs746374229 Chr.2: 111809029-111809029 on Build GRCh38
rs978905940 Chr.2: 111809001-111809001 on Build GRCh38

View Full Product Details