Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201704904-201705177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALS2
Entrez Gene ID
57679
Gene Name
ALS2, alsin Rho guanine nucleotide exchange factor
Gene Aliases
ALS2CR6, ALSJ, IAHSP, PLSJ
UniGene
Hs.471096
Gene Chromosome Location
Chr.2: 201700263-201781172 on Build GRCh38
Transcripts
NM_020919.3 XM_006712654.2 XM_006712655.3 XM_011511530.2 XM_017004569.1 XM_017004571.1 XM_017004572.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
719756 c.4657G>A p.A1553T Chr.2: 201705170-201705170 on Build GRCh38
367290 c.4661C>G p.S1554* Chr.2: 201705166-201705166 on Build GRCh38
7096811 c.4661C>T p.S1554L Chr.2: 201705166-201705166 on Build GRCh38
6466834 c.4688+10G>T p.? Chr.2: 201705129-201705129 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs994900737 Chr.2: 201704946-201704946 on Build GRCh38
rs957274906 Chr.2: 201705049-201705049 on Build GRCh38
rs377461652 Chr.2: 201704969-201704969 on Build GRCh38
rs996518498 Chr.2: 201705128-201705128 on Build GRCh38
rs751152776 Chr.2: 201705036-201705036 on Build GRCh38
rs371130591 Chr.2: 201705069-201705069 on Build GRCh38
rs932493342 Chr.2: 201705078-201705078 on Build GRCh38
rs958860902 Chr.2: 201704973-201704973 on Build GRCh38
rs764067612 Chr.2: 201705117-201705117 on Build GRCh38
rs1024556178 Chr.2: 201705004-201705004 on Build GRCh38
rs776086130 Chr.2: 201705061-201705061 on Build GRCh38
rs758632562 Chr.2: 201705098-201705098 on Build GRCh38
rs977069192 Chr.2: 201705017-201705017 on Build GRCh38
rs989893114 Chr.2: 201705053-201705053 on Build GRCh38
rs1056422553 Chr.2: 201705026-201705026 on Build GRCh38
rs538708500 Chr.2: 201705041-201705041 on Build GRCh38
rs986634433 Chr.2: 201705110-201705110 on Build GRCh38
rs546493164 Chr.2: 201705077-201705077 on Build GRCh38
rs139919006 Chr.2: 201705124-201705124 on Build GRCh38
rs921017248 Chr.2: 201705067-201705067 on Build GRCh38
rs939266758 Chr.2: 201704977-201704977 on Build GRCh38
rs924302544 Chr.2: 201705035-201705035 on Build GRCh38
rs747080250 Chr.2: 201705113-201705113 on Build GRCh38
rs1013511221 Chr.2: 201704974-201704974 on Build GRCh38
rs912461048 Chr.2: 201705119-201705119 on Build GRCh38
rs7598082 Chr.2: 201704931-201704931 on Build GRCh38
rs778145213 Chr.2: 201705111-201705111 on Build GRCh38
rs374097931 Chr.2: 201704970-201704970 on Build GRCh38
rs971681155 Chr.2: 201705014-201705014 on Build GRCh38

View Full Product Details