Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133547149-133547382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL2
Entrez Gene ID
9719
Gene Name
ADAMTS like 2
Gene Aliases
GPHYSD1
UniGene
Hs.522543
Gene Chromosome Location
Chr.9: 133532164-133575519 on Build GRCh38
Transcripts
NM_001145320.1 NM_014694.3 XM_005272237.3 XM_005272238.3 XM_005272239.2 XM_011519241.2 XM_011519242.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5417839 c.1224C>A p.Y408* Chr.9: 133547171-133547171 on Build GRCh38
7329082 c.1257G>A p.L419L Chr.9: 133547204-133547204 on Build GRCh38
5417838 c.897C>A p.Y299* Chr.9: 133547171-133547171 on Build GRCh38
3763725 c.900C>T p.I300I Chr.9: 133547174-133547174 on Build GRCh38
3763726 c.1227C>T p.I409I Chr.9: 133547174-133547174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919516120 Chr.9: 133547203-133547203 on Build GRCh38
rs771566800 Chr.9: 133547241-133547241 on Build GRCh38
rs2073877 Chr.9: 133547174-133547174 on Build GRCh38
rs777881456 Chr.9: 133547194-133547194 on Build GRCh38
rs2073878 Chr.9: 133547224-133547224 on Build GRCh38
rs747215901 Chr.9: 133547198-133547198 on Build GRCh38
rs752724301 Chr.9: 133547175-133547175 on Build GRCh38
rs368709427 Chr.9: 133547237-133547237 on Build GRCh38
rs755230179 Chr.9: 133547228-133547228 on Build GRCh38
rs952218721 Chr.9: 133547342-133547342 on Build GRCh38
rs759912686 Chr.9: 133547249-133547249 on Build GRCh38
rs959490689 Chr.9: 133547348-133547348 on Build GRCh38
rs777130225 Chr.9: 133547247-133547247 on Build GRCh38
rs779098356 Chr.9: 133547231-133547247 on Build GRCh38
rs758446604 Chr.9: 133547186-133547186 on Build GRCh38
rs755905285 Chr.9: 133547202-133547202 on Build GRCh38
rs1003388507 Chr.9: 133547325-133547325 on Build GRCh38
rs765852298 Chr.9: 133547254-133547254 on Build GRCh38
rs779804173 Chr.9: 133547217-133547217 on Build GRCh38
rs752829560 Chr.9: 133547253-133547261 on Build GRCh38
rs577092714 Chr.9: 133547287-133547287 on Build GRCh38
rs373866266 Chr.9: 133547245-133547245 on Build GRCh38
rs936600520 Chr.9: 133547296-133547296 on Build GRCh38
rs372427541 Chr.9: 133547223-133547223 on Build GRCh38
rs753199351 Chr.9: 133547256-133547256 on Build GRCh38
rs1010998598 Chr.9: 133547299-133547299 on Build GRCh38
rs893551305 Chr.9: 133547298-133547298 on Build GRCh38
rs1044880816 Chr.9: 133547284-133547284 on Build GRCh38
rs577332470 Chr.9: 133547361-133547361 on Build GRCh38
rs899585402 Chr.9: 133547322-133547322 on Build GRCh38
rs552622691 Chr.9: 133547343-133547343 on Build GRCh38
rs774432357 Chr.9: 133547201-133547201 on Build GRCh38
rs555262449 Chr.9: 133547289-133547289 on Build GRCh38
rs567082302 Chr.9: 133547304-133547304 on Build GRCh38
rs550989356 Chr.9: 133547240-133547240 on Build GRCh38
rs2073879 Chr.9: 133547260-133547260 on Build GRCh38
rs760349900 Chr.9: 133547243-133547243 on Build GRCh38
rs1020538539 Chr.9: 133547303-133547303 on Build GRCh38
rs764203827 Chr.9: 133547261-133547261 on Build GRCh38
rs2073880 Chr.9: 133547332-133547332 on Build GRCh38
rs533716793 Chr.9: 133547352-133547352 on Build GRCh38

View Full Product Details